Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17822894-17823129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL8
Entrez Gene ID
81569
Gene Name
actin like 8
Gene Aliases
CT57
UniGene
Hs.2149
Gene Chromosome Location
Chr.1: 17755313-17827063 on Build GRCh38
Transcripts
NM_030812.2 XM_011542212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900515 c.96C>T p.I32I Chr.1: 17823104-17823104 on Build GRCh38
216986 c.34T>C p.S12P Chr.1: 17823042-17823042 on Build GRCh38
900516 c.117G>A p.K39K Chr.1: 17823125-17823125 on Build GRCh38
6122936 c.111G>T p.P37P Chr.1: 17823119-17823119 on Build GRCh38
1639638 c.111G>A p.P37P Chr.1: 17823119-17823119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977155836 Chr.1: 17823031-17823031 on Build GRCh38
rs370617672 Chr.1: 17823006-17823006 on Build GRCh38
rs368715674 Chr.1: 17823079-17823079 on Build GRCh38
rs561909733 Chr.1: 17823042-17823042 on Build GRCh38
rs770919704 Chr.1: 17823013-17823013 on Build GRCh38
rs757837876 Chr.1: 17823059-17823059 on Build GRCh38
rs202242597 Chr.1: 17822980-17822980 on Build GRCh38
rs760974264 Chr.1: 17823038-17823038 on Build GRCh38
rs965839676 Chr.1: 17822931-17822931 on Build GRCh38
rs377529807 Chr.1: 17822983-17822983 on Build GRCh38
rs370422420 Chr.1: 17823045-17823045 on Build GRCh38
rs750983259 Chr.1: 17823064-17823064 on Build GRCh38
rs694214 Chr.1: 17823015-17823015 on Build GRCh38
rs183678603 Chr.1: 17823005-17823005 on Build GRCh38
rs12408385 Chr.1: 17823098-17823098 on Build GRCh38
rs754348930 Chr.1: 17822976-17822976 on Build GRCh38
rs371868707 Chr.1: 17823090-17823090 on Build GRCh38
rs774398798 Chr.1: 17823014-17823014 on Build GRCh38
rs755362470 Chr.1: 17823109-17823110 on Build GRCh38
rs774216868 Chr.1: 17823104-17823104 on Build GRCh38
rs2296035 Chr.1: 17823071-17823071 on Build GRCh38
rs747728884 Chr.1: 17823069-17823069 on Build GRCh38
rs773193212 Chr.1: 17823024-17823024 on Build GRCh38
rs1014735883 Chr.1: 17822928-17822928 on Build GRCh38
rs778230710 Chr.1: 17822970-17822970 on Build GRCh38
rs928224943 Chr.1: 17823109-17823109 on Build GRCh38
rs762842483 Chr.1: 17823008-17823008 on Build GRCh38
rs754209555 Chr.1: 17822963-17822964 on Build GRCh38
rs56082318 Chr.1: 17822981-17822981 on Build GRCh38
rs754383232 Chr.1: 17823065-17823065 on Build GRCh38
rs374320130 Chr.1: 17823023-17823023 on Build GRCh38
rs559937741 Chr.1: 17822985-17822985 on Build GRCh38
rs747846271 Chr.1: 17822988-17822988 on Build GRCh38
rs368004903 Chr.1: 17823097-17823097 on Build GRCh38
rs765904980 Chr.1: 17823063-17823063 on Build GRCh38
rs749150517 Chr.1: 17823075-17823075 on Build GRCh38
rs116433945 Chr.1: 17823068-17823068 on Build GRCh38

View Full Product Details