Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75728452-75728677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912144 c.86G>A p.R29Q Chr.1: 75728456-75728456 on Build GRCh38
1667642 c.115T>G p.F39V Chr.1: 75728485-75728485 on Build GRCh38
5516161 c.85C>T p.R29* Chr.1: 75728455-75728455 on Build GRCh38
5516160 c.97C>T p.R33* Chr.1: 75728455-75728455 on Build GRCh38
6591687 c.98G>A p.R33Q Chr.1: 75728456-75728456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754086774 Chr.1: 75728505-75728505 on Build GRCh38
rs890094593 Chr.1: 75728583-75728583 on Build GRCh38
rs1029880524 Chr.1: 75728568-75728568 on Build GRCh38
rs933400092 Chr.1: 75728593-75728593 on Build GRCh38
rs533818662 Chr.1: 75728588-75728588 on Build GRCh38
rs875989868 Chr.1: 75728602-75728602 on Build GRCh38
rs774074624 Chr.1: 75728476-75728476 on Build GRCh38
rs765332322 Chr.1: 75728519-75728519 on Build GRCh38
rs758755552 Chr.1: 75728525-75728525 on Build GRCh38
rs113754652 Chr.1: 75728490-75728490 on Build GRCh38
rs202020471 Chr.1: 75728549-75728549 on Build GRCh38
rs773396853 Chr.1: 75728610-75728610 on Build GRCh38
rs200374325 Chr.1: 75728526-75728526 on Build GRCh38
rs904369281 Chr.1: 75728628-75728628 on Build GRCh38
rs772586266 Chr.1: 75728484-75728484 on Build GRCh38
rs796117827 Chr.1: 75728548-75728548 on Build GRCh38
rs752929464 Chr.1: 75728522-75728522 on Build GRCh38
rs755405418 Chr.1: 75728506-75728506 on Build GRCh38
rs772778486 Chr.1: 75728495-75728495 on Build GRCh38
rs398123071 Chr.1: 75728488-75728488 on Build GRCh38
rs113887538 Chr.1: 75728489-75728489 on Build GRCh38
rs548538699 Chr.1: 75728496-75728496 on Build GRCh38
rs1146572 Chr.1: 75728621-75728621 on Build GRCh38
rs1051797979 Chr.1: 75728594-75728594 on Build GRCh38
rs75993895 Chr.1: 75728545-75728545 on Build GRCh38
rs875989853 Chr.1: 75728550-75728550 on Build GRCh38
rs377330041 Chr.1: 75728535-75728535 on Build GRCh38
rs202218503 Chr.1: 75728498-75728498 on Build GRCh38
rs766711158 Chr.1: 75728637-75728637 on Build GRCh38
rs756047234 Chr.1: 75728531-75728531 on Build GRCh38
rs875989875 Chr.1: 75728483-75728484 on Build GRCh38
rs998732625 Chr.1: 75728634-75728634 on Build GRCh38
rs1006082856 Chr.1: 75728483-75728483 on Build GRCh38
rs991381916 Chr.1: 75728628-75728629 on Build GRCh38
rs552153704 Chr.1: 75728612-75728612 on Build GRCh38
rs889104731 Chr.1: 75728607-75728607 on Build GRCh38
rs1008518786 Chr.1: 75728584-75728584 on Build GRCh38
rs761421407 Chr.1: 75728492-75728492 on Build GRCh38
rs958799641 Chr.1: 75728627-75728627 on Build GRCh38
rs747096320 Chr.1: 75728529-75728529 on Build GRCh38

View Full Product Details