Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 100532092-100532361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGACT
Entrez Gene ID
87769
Gene Name
gamma-glutamylamine cyclotransferase
Gene Aliases
A2LD1
UniGene
Hs.350868
Gene Chromosome Location
Chr.13: 100530164-100588792 on Build GRCh38
Transcripts
NM_001195087.1 NM_033110.2 XM_005254083.2 XM_011521129.2 XM_017020804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545457 c.378G>A p.P126P Chr.13: 100532214-100532214 on Build GRCh38
4856401 c.294G>A p.L98L Chr.13: 100532298-100532298 on Build GRCh38
944993 c.243C>T p.F81F Chr.13: 100532349-100532349 on Build GRCh38
5486865 c.309G>A p.P103P Chr.13: 100532283-100532283 on Build GRCh38
5628242 c.292C>A p.L98M Chr.13: 100532300-100532300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940089446 Chr.13: 100532204-100532204 on Build GRCh38
rs530900700 Chr.13: 100532201-100532201 on Build GRCh38
rs545940682 Chr.13: 100532224-100532224 on Build GRCh38
rs1037573033 Chr.13: 100532310-100532310 on Build GRCh38
rs1008132649 Chr.13: 100532298-100532298 on Build GRCh38
rs369036128 Chr.13: 100532251-100532251 on Build GRCh38
rs749568349 Chr.13: 100532319-100532319 on Build GRCh38
rs781488724 Chr.13: 100532294-100532294 on Build GRCh38
rs377460144 Chr.13: 100532196-100532196 on Build GRCh38
rs373840929 Chr.13: 100532122-100532122 on Build GRCh38
rs775817783 Chr.13: 100532268-100532269 on Build GRCh38
rs886360281 Chr.13: 100532297-100532297 on Build GRCh38
rs1001724757 Chr.13: 100532166-100532166 on Build GRCh38
rs755844327 Chr.13: 100532197-100532197 on Build GRCh38
rs926868561 Chr.13: 100532155-100532155 on Build GRCh38
rs747598598 Chr.13: 100532278-100532278 on Build GRCh38
rs766605344 Chr.13: 100532328-100532328 on Build GRCh38
rs553670551 Chr.13: 100532139-100532139 on Build GRCh38
rs770236696 Chr.13: 100532312-100532312 on Build GRCh38
rs61747479 Chr.13: 100532167-100532167 on Build GRCh38
rs978763139 Chr.13: 100532215-100532215 on Build GRCh38
rs757853458 Chr.13: 100532289-100532289 on Build GRCh38
rs908746136 Chr.13: 100532171-100532171 on Build GRCh38
rs933159872 Chr.13: 100532269-100532269 on Build GRCh38
rs922695581 Chr.13: 100532268-100532268 on Build GRCh38
rs563916503 Chr.13: 100532168-100532168 on Build GRCh38
rs898478330 Chr.13: 100532235-100532235 on Build GRCh38
rs564118954 Chr.13: 100532313-100532313 on Build GRCh38
rs192075410 Chr.13: 100532156-100532156 on Build GRCh38
rs746251971 Chr.13: 100532309-100532309 on Build GRCh38
rs866187114 Chr.13: 100532182-100532182 on Build GRCh38
rs1004723113 Chr.13: 100532300-100532300 on Build GRCh38
rs967601270 Chr.13: 100532213-100532213 on Build GRCh38
rs980989467 Chr.13: 100532146-100532146 on Build GRCh38
rs772210182 Chr.13: 100532154-100532155 on Build GRCh38
rs963105712 Chr.13: 100532160-100532160 on Build GRCh38
rs932668497 Chr.13: 100532286-100532286 on Build GRCh38
rs942672053 Chr.13: 100532229-100532229 on Build GRCh38
rs1040214394 Chr.13: 100532263-100532263 on Build GRCh38
rs891039111 Chr.13: 100532290-100532290 on Build GRCh38
rs188503423 Chr.13: 100532340-100532340 on Build GRCh38
rs768743482 Chr.13: 100532331-100532331 on Build GRCh38
rs1051580229 Chr.13: 100532288-100532288 on Build GRCh38
rs183442395 Chr.13: 100532205-100532205 on Build GRCh38
rs1049599577 Chr.13: 100532287-100532287 on Build GRCh38
rs774553677 Chr.13: 100532338-100532338 on Build GRCh38
rs1021688999 Chr.13: 100532136-100532136 on Build GRCh38

View Full Product Details