Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74290322-74290549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
UniGene
Hs.94395
Gene Chromosome Location
Chr.14: 74285277-74303064 on Build GRCh38
Transcripts
NM_005050.3 NR_003256.2 XM_005267939.3 XM_005267940.3 XM_005267941.4 XM_005267942.4 XM_005267946.3 XM_005267949.3 XM_006720223.2 XM_011537041.2 XM_017021531.1 XM_017021532.1 XM_017021533.1 XM_017021534.1 XM_017021535.1 XM_017021536.1 XM_017021537.1 XM_017021538.1 XM_017021539.1 XM_017021540.1 XM_017021541.1 XM_017021542.1 XM_017021543.1 XM_017021544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6317287 c.1263C>A p.G421G Chr.14: 74290355-74290355 on Build GRCh38
4460910 c.1184C>T p.S395F Chr.14: 74290434-74290434 on Build GRCh38
470230 c.1216G>C p.D406H Chr.14: 74290402-74290402 on Build GRCh38
1678129 c.1171C>T p.R391W Chr.14: 74290447-74290447 on Build GRCh38
234341 c.1164C>T p.L388L Chr.14: 74290454-74290454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768330414 Chr.14: 74290344-74290344 on Build GRCh38
rs369452408 Chr.14: 74290522-74290522 on Build GRCh38
rs868292947 Chr.14: 74290438-74290438 on Build GRCh38
rs756862834 Chr.14: 74290411-74290411 on Build GRCh38
rs776821878 Chr.14: 74290490-74290490 on Build GRCh38
rs747987460 Chr.14: 74290446-74290446 on Build GRCh38
rs775236037 Chr.14: 74290523-74290524 on Build GRCh38
rs765567501 Chr.14: 74290405-74290405 on Build GRCh38
rs780846726 Chr.14: 74290414-74290414 on Build GRCh38
rs530167374 Chr.14: 74290527-74290527 on Build GRCh38
rs773345921 Chr.14: 74290371-74290372 on Build GRCh38
rs755569781 Chr.14: 74290402-74290402 on Build GRCh38
rs770903927 Chr.14: 74290485-74290485 on Build GRCh38
rs774256579 Chr.14: 74290439-74290439 on Build GRCh38
rs745327264 Chr.14: 74290419-74290419 on Build GRCh38
rs993168511 Chr.14: 74290348-74290348 on Build GRCh38
rs941046002 Chr.14: 74290494-74290494 on Build GRCh38
rs767492476 Chr.14: 74290501-74290502 on Build GRCh38
rs1037097519 Chr.14: 74290500-74290500 on Build GRCh38
rs145155300 Chr.14: 74290388-74290388 on Build GRCh38
rs773381798 Chr.14: 74290371-74290371 on Build GRCh38
rs988847476 Chr.14: 74290510-74290510 on Build GRCh38
rs1026011443 Chr.14: 74290357-74290357 on Build GRCh38
rs749168282 Chr.14: 74290428-74290428 on Build GRCh38
rs759420952 Chr.14: 74290464-74290464 on Build GRCh38
rs765765487 Chr.14: 74290507-74290507 on Build GRCh38
rs965299774 Chr.14: 74290427-74290427 on Build GRCh38
rs753015601 Chr.14: 74290521-74290521 on Build GRCh38
rs753105481 Chr.14: 74290407-74290407 on Build GRCh38
rs988025549 Chr.14: 74290422-74290422 on Build GRCh38
rs373646439 Chr.14: 74290523-74290523 on Build GRCh38
rs201164224 Chr.14: 74290382-74290382 on Build GRCh38
rs774344504 Chr.14: 74290349-74290349 on Build GRCh38
rs773580031 Chr.14: 74290469-74290469 on Build GRCh38
rs759929736 Chr.14: 74290496-74290496 on Build GRCh38
rs762104370 Chr.14: 74290364-74290364 on Build GRCh38
rs149139488 Chr.14: 74290482-74290482 on Build GRCh38
rs376867854 Chr.14: 74290528-74290528 on Build GRCh38
rs779680204 Chr.14: 74290425-74290425 on Build GRCh38
rs374984401 Chr.14: 74290381-74290381 on Build GRCh38
rs372941783 Chr.14: 74290366-74290366 on Build GRCh38
rs772370453 Chr.14: 74290455-74290455 on Build GRCh38
rs768762298 Chr.14: 74290436-74290436 on Build GRCh38
rs755586441 Chr.14: 74290424-74290424 on Build GRCh38
rs979042752 Chr.14: 74290478-74290478 on Build GRCh38

View Full Product Details