Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63835161-63835432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf49
Entrez Gene ID
132200
Gene Name
chromosome 3 open reading frame 49
Gene Aliases
-
UniGene
Hs.506386
Gene Chromosome Location
Chr.3: 63819365-63848636 on Build GRCh38
Transcripts
NR_026866.1 NM_001285387.2 NM_001285404.1 NM_025075.3 XM_006713339.3

Gene Symbol
THOC7
Entrez Gene ID
80145
Gene Name
THO complex 7
Gene Aliases
NIF3L1BP1, fSAP24, hTREX30
UniGene
Hs.288151
Gene Chromosome Location
Chr.3: 63833870-63864478 on Build GRCh38
Transcripts
NR_026866.1 NM_001285387.2 NM_001285404.1 NM_025075.3 XM_006713339.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84808 c.478-3C>A p.? Chr.3: 63835226-63835226 on Build GRCh38
6310490 c.522C>A p.I174I Chr.3: 63835179-63835179 on Build GRCh38
1048003 c.503A>G p.H168R Chr.3: 63835198-63835198 on Build GRCh38
5044631 c.417A>T p.L139L Chr.3: 63835384-63835384 on Build GRCh38
6317979 c.453C>A p.H151Q Chr.3: 63835348-63835348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774210324 Chr.3: 63835255-63835255 on Build GRCh38
rs573190557 Chr.3: 63835252-63835252 on Build GRCh38
rs529873264 Chr.3: 63835371-63835371 on Build GRCh38
rs759751230 Chr.3: 63835318-63835318 on Build GRCh38
rs759005934 Chr.3: 63835338-63835338 on Build GRCh38
rs747215872 Chr.3: 63835194-63835194 on Build GRCh38
rs373470489 Chr.3: 63835287-63835287 on Build GRCh38
rs370012958 Chr.3: 63835275-63835275 on Build GRCh38
rs375558290 Chr.3: 63835213-63835213 on Build GRCh38
rs749027288 Chr.3: 63835228-63835228 on Build GRCh38
rs776123092 Chr.3: 63835266-63835269 on Build GRCh38
rs765934313 Chr.3: 63835313-63835313 on Build GRCh38
rs774042036 Chr.3: 63835284-63835287 on Build GRCh38
rs574808987 Chr.3: 63835302-63835302 on Build GRCh38
rs771526976 Chr.3: 63835291-63835291 on Build GRCh38
rs751573300 Chr.3: 63835345-63835345 on Build GRCh38
rs773259429 Chr.3: 63835372-63835372 on Build GRCh38
rs62253803 Chr.3: 63835407-63835407 on Build GRCh38
rs202003417 Chr.3: 63835317-63835317 on Build GRCh38
rs140289523 Chr.3: 63835329-63835329 on Build GRCh38
rs1051050841 Chr.3: 63835365-63835365 on Build GRCh38
rs949667052 Chr.3: 63835217-63835217 on Build GRCh38
rs562345461 Chr.3: 63835357-63835357 on Build GRCh38
rs746495156 Chr.3: 63835398-63835398 on Build GRCh38
rs777002257 Chr.3: 63835288-63835291 on Build GRCh38
rs373519986 Chr.3: 63835205-63835205 on Build GRCh38
rs764738747 Chr.3: 63835344-63835344 on Build GRCh38
rs747974118 Chr.3: 63835366-63835366 on Build GRCh38
rs137931328 Chr.3: 63835346-63835346 on Build GRCh38
rs778707914 Chr.3: 63835364-63835364 on Build GRCh38
rs754716011 Chr.3: 63835359-63835359 on Build GRCh38
rs540211076 Chr.3: 63835267-63835267 on Build GRCh38
rs771028360 Chr.3: 63835197-63835197 on Build GRCh38
rs753208450 Chr.3: 63835337-63835337 on Build GRCh38
rs1047055877 Chr.3: 63835261-63835261 on Build GRCh38
rs772674423 Chr.3: 63835297-63835297 on Build GRCh38
rs574009018 Chr.3: 63835281-63835282 on Build GRCh38
rs915397807 Chr.3: 63835298-63835298 on Build GRCh38
rs1015438131 Chr.3: 63835390-63835390 on Build GRCh38
rs775302763 Chr.3: 63835387-63835387 on Build GRCh38

View Full Product Details