Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203165421-203165684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA1
Entrez Gene ID
134
Gene Name
adenosine A1 receptor
Gene Aliases
RDC7
UniGene
Hs.77867
Gene Chromosome Location
Chr.1: 203127705-203167405 on Build GRCh38
Transcripts
NM_000674.2 NM_001048230.1 XM_005244901.1 XM_005244902.4 XM_017000342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595313 c.744G>C p.L248L Chr.1: 203165663-203165663 on Build GRCh38
6595315 c.536A>G p.Y179C Chr.1: 203165455-203165455 on Build GRCh38
2213515 c.623G>A p.R208H Chr.1: 203165542-203165542 on Build GRCh38
5451203 c.648G>A p.S216S Chr.1: 203165567-203165567 on Build GRCh38
2213517 c.737G>C p.S246T Chr.1: 203165656-203165656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778641521 Chr.1: 203165580-203165580 on Build GRCh38
rs539069853 Chr.1: 203165566-203165566 on Build GRCh38
rs1033168113 Chr.1: 203165553-203165553 on Build GRCh38
rs764447031 Chr.1: 203165543-203165543 on Build GRCh38
rs144484179 Chr.1: 203165520-203165520 on Build GRCh38
rs751801919 Chr.1: 203165552-203165552 on Build GRCh38
rs745321082 Chr.1: 203165464-203165464 on Build GRCh38
rs762703952 Chr.1: 203165527-203165529 on Build GRCh38
rs537966964 Chr.1: 203165624-203165624 on Build GRCh38
rs768403618 Chr.1: 203165494-203165494 on Build GRCh38
rs779515290 Chr.1: 203165570-203165570 on Build GRCh38
rs769965850 Chr.1: 203165534-203165534 on Build GRCh38
rs74912377 Chr.1: 203165504-203165504 on Build GRCh38
rs767961013 Chr.1: 203165563-203165563 on Build GRCh38
rs201016113 Chr.1: 203165576-203165576 on Build GRCh38
rs199516537 Chr.1: 203165661-203165661 on Build GRCh38
rs747980090 Chr.1: 203165501-203165501 on Build GRCh38
rs375943917 Chr.1: 203165480-203165480 on Build GRCh38
rs1019641966 Chr.1: 203165505-203165505 on Build GRCh38
rs373232724 Chr.1: 203165487-203165487 on Build GRCh38
rs746924602 Chr.1: 203165600-203165600 on Build GRCh38
rs146573702 Chr.1: 203165633-203165633 on Build GRCh38
rs937429758 Chr.1: 203165477-203165477 on Build GRCh38
rs139946190 Chr.1: 203165542-203165542 on Build GRCh38
rs368740274 Chr.1: 203165490-203165490 on Build GRCh38
rs200239809 Chr.1: 203165567-203165567 on Build GRCh38
rs148415221 Chr.1: 203165541-203165541 on Build GRCh38
rs201445170 Chr.1: 203165495-203165495 on Build GRCh38
rs774651930 Chr.1: 203165628-203165628 on Build GRCh38
rs199949273 Chr.1: 203165502-203165502 on Build GRCh38
rs754581480 Chr.1: 203165577-203165577 on Build GRCh38
rs200961926 Chr.1: 203165489-203165489 on Build GRCh38
rs75070458 Chr.1: 203165554-203165554 on Build GRCh38
rs775089272 Chr.1: 203165447-203165455 on Build GRCh38
rs775712225 Chr.1: 203165619-203165619 on Build GRCh38
rs777752008 Chr.1: 203165597-203165597 on Build GRCh38
rs747849534 Chr.1: 203165584-203165584 on Build GRCh38
rs201849915 Chr.1: 203165585-203165585 on Build GRCh38
rs201570528 Chr.1: 203165511-203165511 on Build GRCh38
rs773306244 Chr.1: 203165509-203165509 on Build GRCh38
rs200281933 Chr.1: 203165615-203165615 on Build GRCh38
rs781665257 Chr.1: 203165451-203165451 on Build GRCh38
rs201083386 Chr.1: 203165623-203165623 on Build GRCh38

View Full Product Details