Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73593817-73594042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT4
Entrez Gene ID
122970
Gene Name
acyl-CoA thioesterase 4
Gene Aliases
PTE-Ib, PTE1B, PTE2B
UniGene
Hs.49433
Gene Chromosome Location
Chr.14: 73591528-73595766 on Build GRCh38
Transcripts
NM_152331.3 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NM_152331.3 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940416 c.611C>T p.S204F Chr.14: 73593855-73593855 on Build GRCh38
6328159 c.660+9C>A p.? Chr.14: 73593913-73593913 on Build GRCh38
698703 c.612C>T p.S204S Chr.14: 73593856-73593856 on Build GRCh38
4630080 c.596A>C p.N199T Chr.14: 73593840-73593840 on Build GRCh38
5722228 c.660+2T>C p.? Chr.14: 73593906-73593906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992978394 Chr.14: 73593974-73593974 on Build GRCh38
rs767883379 Chr.14: 73593851-73593851 on Build GRCh38
rs966904670 Chr.14: 73593944-73593944 on Build GRCh38
rs201663408 Chr.14: 73593869-73593869 on Build GRCh38
rs750676294 Chr.14: 73593945-73593945 on Build GRCh38
rs889907727 Chr.14: 73594012-73594012 on Build GRCh38
rs781086824 Chr.14: 73593936-73593936 on Build GRCh38
rs369066813 Chr.14: 73593975-73593975 on Build GRCh38
rs187102550 Chr.14: 73593888-73593888 on Build GRCh38
rs1042375728 Chr.14: 73593914-73593914 on Build GRCh38
rs372772376 Chr.14: 73593882-73593882 on Build GRCh38
rs751671920 Chr.14: 73593852-73593852 on Build GRCh38
rs776366966 Chr.14: 73593899-73593899 on Build GRCh38
rs369330963 Chr.14: 73593915-73593915 on Build GRCh38
rs574322158 Chr.14: 73593918-73593918 on Build GRCh38
rs202000176 Chr.14: 73593880-73593880 on Build GRCh38
rs765085379 Chr.14: 73593905-73593905 on Build GRCh38
rs1051599976 Chr.14: 73593972-73593972 on Build GRCh38
rs770305203 Chr.14: 73593895-73593895 on Build GRCh38
rs915700245 Chr.14: 73593993-73593993 on Build GRCh38
rs770219539 Chr.14: 73593877-73593877 on Build GRCh38
rs35909515 Chr.14: 73593868-73593868 on Build GRCh38
rs759052366 Chr.14: 73593901-73593901 on Build GRCh38
rs780342168 Chr.14: 73593878-73593878 on Build GRCh38
rs978330218 Chr.14: 73593955-73593955 on Build GRCh38
rs751726829 Chr.14: 73593926-73593926 on Build GRCh38
rs76715250 Chr.14: 73593845-73593845 on Build GRCh38
rs757392851 Chr.14: 73593932-73593932 on Build GRCh38
rs760212540 Chr.14: 73593889-73593889 on Build GRCh38
rs938356375 Chr.14: 73593969-73593969 on Build GRCh38
rs934048961 Chr.14: 73593953-73593953 on Build GRCh38
rs924239790 Chr.14: 73593957-73593957 on Build GRCh38
rs746040937 Chr.14: 73593873-73593873 on Build GRCh38

View Full Product Details