Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110112714-110112981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
UniGene
Hs.501012
Gene Chromosome Location
Chr.10: 109996350-110135565 on Build GRCh38
Transcripts
NM_001121.3 NM_001320591.1 NM_001320592.1 NM_001320593.1 NM_001320594.1 NM_016824.4 NM_019903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6064649 c.202C>T p.R68W Chr.10: 110112783-110112783 on Build GRCh38
3434082 c.198C>T p.A66A Chr.10: 110112779-110112779 on Build GRCh38
4011540 c.245G>A p.G82D Chr.10: 110112826-110112826 on Build GRCh38
2061045 c.332T>G p.L111R Chr.10: 110112913-110112913 on Build GRCh38
6595156 c.203G>A p.R68Q Chr.10: 110112784-110112784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147859836 Chr.10: 110112893-110112893 on Build GRCh38
rs371961813 Chr.10: 110112826-110112826 on Build GRCh38
rs754356569 Chr.10: 110112784-110112784 on Build GRCh38
rs755609771 Chr.10: 110112905-110112905 on Build GRCh38
rs779189030 Chr.10: 110112829-110112829 on Build GRCh38
rs772374757 Chr.10: 110112854-110112854 on Build GRCh38
rs753821732 Chr.10: 110112944-110112944 on Build GRCh38
rs765637788 Chr.10: 110112883-110112883 on Build GRCh38
rs748417464 Chr.10: 110112848-110112848 on Build GRCh38
rs749885468 Chr.10: 110112799-110112799 on Build GRCh38
rs914623953 Chr.10: 110112783-110112783 on Build GRCh38
rs771612215 Chr.10: 110112862-110112862 on Build GRCh38
rs955336473 Chr.10: 110112873-110112873 on Build GRCh38
rs754816683 Chr.10: 110112945-110112945 on Build GRCh38
rs920453861 Chr.10: 110112919-110112919 on Build GRCh38
rs139343347 Chr.10: 110112892-110112892 on Build GRCh38
rs777179853 Chr.10: 110112863-110112863 on Build GRCh38
rs957462600 Chr.10: 110112814-110112814 on Build GRCh38
rs754190904 Chr.10: 110112745-110112745 on Build GRCh38
rs747051168 Chr.10: 110112859-110112859 on Build GRCh38
rs780019495 Chr.10: 110112813-110112813 on Build GRCh38
rs766296422 Chr.10: 110112914-110112914 on Build GRCh38
rs756834111 Chr.10: 110112779-110112779 on Build GRCh38
rs759882827 Chr.10: 110112880-110112880 on Build GRCh38
rs747915497 Chr.10: 110112953-110112953 on Build GRCh38
rs780693571 Chr.10: 110112794-110112794 on Build GRCh38
rs778508199 Chr.10: 110112843-110112843 on Build GRCh38
rs759920339 Chr.10: 110112750-110112750 on Build GRCh38
rs773453290 Chr.10: 110112858-110112858 on Build GRCh38
rs780602898 Chr.10: 110112923-110112923 on Build GRCh38
rs41291886 Chr.10: 110112885-110112885 on Build GRCh38
rs751218662 Chr.10: 110112774-110112774 on Build GRCh38
rs749097779 Chr.10: 110112817-110112817 on Build GRCh38
rs999009094 Chr.10: 110112753-110112753 on Build GRCh38
rs186032453 Chr.10: 110112760-110112760 on Build GRCh38
rs537446217 Chr.10: 110112809-110112809 on Build GRCh38
rs776758541 Chr.10: 110112940-110112941 on Build GRCh38
rs761476337 Chr.10: 110112886-110112886 on Build GRCh38

View Full Product Details