Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50892117-50892386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADNP
Entrez Gene ID
23394
Gene Name
activity dependent neuroprotector homeobox
Gene Aliases
ADNP1, HVDAS, MRD28
UniGene
Hs.293736
Gene Chromosome Location
Chr.20: 50888918-50934984 on Build GRCh38
Transcripts
NM_001282531.1 NM_001282532.1 NM_015339.3 NM_181442.2 XM_011528747.2 XM_011528748.2 XM_017027757.1 XM_017027758.1 XM_017027759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792500 c.2406C>G p.S802S Chr.20: 50892308-50892308 on Build GRCh38
7155852 c.2464G>T p.V822L Chr.20: 50892250-50892250 on Build GRCh38
6450061 c.2387G>A p.W796* Chr.20: 50892327-50892327 on Build GRCh38
287472 c.2453A>G p.Y818C Chr.20: 50892261-50892261 on Build GRCh38
7313206 c.2502C>T p.V834V Chr.20: 50892212-50892212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778752321 Chr.20: 50892147-50892147 on Build GRCh38
rs917063825 Chr.20: 50892217-50892217 on Build GRCh38
rs1005610285 Chr.20: 50892284-50892284 on Build GRCh38
rs998441317 Chr.20: 50892316-50892316 on Build GRCh38
rs766380582 Chr.20: 50892233-50892233 on Build GRCh38
rs779426648 Chr.20: 50892341-50892341 on Build GRCh38
rs146508276 Chr.20: 50892308-50892308 on Build GRCh38
rs144756647 Chr.20: 50892179-50892179 on Build GRCh38
rs200422011 Chr.20: 50892344-50892344 on Build GRCh38
rs587777522 Chr.20: 50892220-50892223 on Build GRCh38
rs77011757 Chr.20: 50892175-50892175 on Build GRCh38
rs587777523 Chr.20: 50892215-50892218 on Build GRCh38
rs148502910 Chr.20: 50892239-50892239 on Build GRCh38
rs111899848 Chr.20: 50892351-50892351 on Build GRCh38
rs200939342 Chr.20: 50892145-50892145 on Build GRCh38
rs753592603 Chr.20: 50892258-50892258 on Build GRCh38
rs765413436 Chr.20: 50892205-50892205 on Build GRCh38
rs777162165 Chr.20: 50892363-50892363 on Build GRCh38
rs776965955 Chr.20: 50892183-50892183 on Build GRCh38
rs757859799 Chr.20: 50892314-50892314 on Build GRCh38
rs527889897 Chr.20: 50892357-50892357 on Build GRCh38
rs755878700 Chr.20: 50892144-50892144 on Build GRCh38
rs753658750 Chr.20: 50892251-50892251 on Build GRCh38
rs888502245 Chr.20: 50892278-50892278 on Build GRCh38
rs752608750 Chr.20: 50892237-50892237 on Build GRCh38
rs369712215 Chr.20: 50892219-50892219 on Build GRCh38
rs773336998 Chr.20: 50892210-50892210 on Build GRCh38
rs776007948 Chr.20: 50892160-50892160 on Build GRCh38
rs779603706 Chr.20: 50892320-50892320 on Build GRCh38
rs745345564 Chr.20: 50892149-50892149 on Build GRCh38
rs757806356 Chr.20: 50892305-50892305 on Build GRCh38
rs897653211 Chr.20: 50892191-50892191 on Build GRCh38
rs1062651 Chr.20: 50892146-50892146 on Build GRCh38
rs755471290 Chr.20: 50892162-50892164 on Build GRCh38
rs762213957 Chr.20: 50892202-50892202 on Build GRCh38
rs149795234 Chr.20: 50892173-50892173 on Build GRCh38
rs778662527 Chr.20: 50892263-50892263 on Build GRCh38
rs376827094 Chr.20: 50892153-50892153 on Build GRCh38
rs940568768 Chr.20: 50892220-50892220 on Build GRCh38
rs142832192 Chr.20: 50892248-50892248 on Build GRCh38
rs886042001 Chr.20: 50892336-50892336 on Build GRCh38

View Full Product Details