Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110041711-110041938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP1
Entrez Gene ID
85369
Gene Name
striatin interacting protein 1
Gene Aliases
FAM40A, FAR11A
UniGene
Hs.584996
Gene Chromosome Location
Chr.1: 110031577-110054641 on Build GRCh38
Transcripts
NM_001270768.1 NM_033088.3 NR_073071.1 XM_006710995.2 XM_017002629.1 XM_017002630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5622520 c.789C>T p.A263A Chr.1: 110041765-110041765 on Build GRCh38
1238836 c.831C>T p.H277H Chr.1: 110041807-110041807 on Build GRCh38
4793694 c.780G>A p.E260E Chr.1: 110041756-110041756 on Build GRCh38
7114666 c.864G>A p.L288L Chr.1: 110041840-110041840 on Build GRCh38
7414372 c.800T>C p.F267S Chr.1: 110041776-110041776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893218628 Chr.1: 110041738-110041738 on Build GRCh38
rs750223219 Chr.1: 110041903-110041903 on Build GRCh38
rs753699475 Chr.1: 110041816-110041816 on Build GRCh38
rs745856445 Chr.1: 110041890-110041890 on Build GRCh38
rs747371053 Chr.1: 110041822-110041822 on Build GRCh38
rs1049215077 Chr.1: 110041835-110041835 on Build GRCh38
rs757192275 Chr.1: 110041862-110041862 on Build GRCh38
rs143991617 Chr.1: 110041777-110041777 on Build GRCh38
rs755986469 Chr.1: 110041907-110041907 on Build GRCh38
rs767279135 Chr.1: 110041899-110041899 on Build GRCh38
rs771431613 Chr.1: 110041740-110041740 on Build GRCh38
rs1014941061 Chr.1: 110041879-110041879 on Build GRCh38
rs765475019 Chr.1: 110041746-110041746 on Build GRCh38
rs760966929 Chr.1: 110041734-110041734 on Build GRCh38
rs751322617 Chr.1: 110041766-110041766 on Build GRCh38
rs761651857 Chr.1: 110041894-110041894 on Build GRCh38
rs768247212 Chr.1: 110041807-110041807 on Build GRCh38
rs774482530 Chr.1: 110041893-110041893 on Build GRCh38
rs759669218 Chr.1: 110041744-110041744 on Build GRCh38
rs752963249 Chr.1: 110041749-110041749 on Build GRCh38
rs146210829 Chr.1: 110041752-110041752 on Build GRCh38
rs888050391 Chr.1: 110041855-110041855 on Build GRCh38
rs777047919 Chr.1: 110041741-110041741 on Build GRCh38
rs781165742 Chr.1: 110041771-110041771 on Build GRCh38
rs923573675 Chr.1: 110041763-110041763 on Build GRCh38
rs766831946 Chr.1: 110041897-110041909 on Build GRCh38
rs763150463 Chr.1: 110041889-110041889 on Build GRCh38
rs372140390 Chr.1: 110041811-110041811 on Build GRCh38
rs771251190 Chr.1: 110041843-110041843 on Build GRCh38
rs755613448 Chr.1: 110041782-110041782 on Build GRCh38
rs376792984 Chr.1: 110041867-110041867 on Build GRCh38
rs775657111 Chr.1: 110041888-110041888 on Build GRCh38
rs757040072 Chr.1: 110041769-110041769 on Build GRCh38
rs748810002 Chr.1: 110041801-110041801 on Build GRCh38
rs777182211 Chr.1: 110041848-110041848 on Build GRCh38
rs900685601 Chr.1: 110041793-110041793 on Build GRCh38
rs779472272 Chr.1: 110041788-110041788 on Build GRCh38
rs201975986 Chr.1: 110041851-110041851 on Build GRCh38
rs764236973 Chr.1: 110041756-110041756 on Build GRCh38
rs996333410 Chr.1: 110041808-110041808 on Build GRCh38

View Full Product Details