Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46719191-46719460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2
Entrez Gene ID
2147
Gene Name
coagulation factor II, thrombin
Gene Aliases
PT, RPRGL2, THPH1
UniGene
Hs.655207
Gene Chromosome Location
Chr.11: 46719166-46739508 on Build GRCh38
Transcripts
NM_000506.4 NM_001311257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986181 c.10G>A p.V4I Chr.11: 46719245-46719245 on Build GRCh38
6276980 c.56G>T p.C19F Chr.11: 46719291-46719291 on Build GRCh38
4033065 c.6G>A p.A2A Chr.11: 46719241-46719241 on Build GRCh38
6068734 c.22C>A p.Q8K Chr.11: 46719257-46719257 on Build GRCh38
325286 c.41C>G p.A14G Chr.11: 46719276-46719276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035565130 Chr.11: 46719395-46719395 on Build GRCh38
rs775021214 Chr.11: 46719248-46719248 on Build GRCh38
rs745611659 Chr.11: 46719213-46719213 on Build GRCh38
rs372560648 Chr.11: 46719324-46719324 on Build GRCh38
rs779814226 Chr.11: 46719329-46719329 on Build GRCh38
rs886048333 Chr.11: 46719280-46719280 on Build GRCh38
rs775929245 Chr.11: 46719364-46719364 on Build GRCh38
rs766767175 Chr.11: 46719313-46719314 on Build GRCh38
rs772718500 Chr.11: 46719359-46719359 on Build GRCh38
rs763600313 Chr.11: 46719374-46719374 on Build GRCh38
rs755345665 Chr.11: 46719314-46719315 on Build GRCh38
rs763098369 Chr.11: 46719244-46719244 on Build GRCh38
rs767987956 Chr.11: 46719264-46719264 on Build GRCh38
rs773515508 Chr.11: 46719347-46719347 on Build GRCh38
rs145969693 Chr.11: 46719357-46719357 on Build GRCh38
rs866155797 Chr.11: 46719219-46719219 on Build GRCh38
rs770938955 Chr.11: 46719316-46719317 on Build GRCh38
rs766756950 Chr.11: 46719245-46719245 on Build GRCh38
rs758400083 Chr.11: 46719327-46719327 on Build GRCh38
rs753243533 Chr.11: 46719309-46719309 on Build GRCh38
rs758038546 Chr.11: 46719249-46719249 on Build GRCh38
rs775306348 Chr.11: 46719240-46719240 on Build GRCh38
rs756810084 Chr.11: 46719319-46719319 on Build GRCh38
rs763811997 Chr.11: 46719285-46719285 on Build GRCh38
rs993553459 Chr.11: 46719280-46719280 on Build GRCh38
rs773112047 Chr.11: 46719242-46719242 on Build GRCh38
rs761409107 Chr.11: 46719434-46719434 on Build GRCh38
rs755663766 Chr.11: 46719274-46719274 on Build GRCh38
rs769845958 Chr.11: 46719331-46719331 on Build GRCh38
rs760170533 Chr.11: 46719263-46719263 on Build GRCh38
rs764725211 Chr.11: 46719435-46719435 on Build GRCh38
rs761128414 Chr.11: 46719368-46719368 on Build GRCh38
rs905125792 Chr.11: 46719340-46719340 on Build GRCh38
rs748368751 Chr.11: 46719241-46719241 on Build GRCh38
rs753503722 Chr.11: 46719402-46719402 on Build GRCh38
rs1051980466 Chr.11: 46719234-46719234 on Build GRCh38
rs771829437 Chr.11: 46719218-46719218 on Build GRCh38
rs753169407 Chr.11: 46719269-46719269 on Build GRCh38
rs753780393 Chr.11: 46719369-46719369 on Build GRCh38
rs749279091 Chr.11: 46719349-46719349 on Build GRCh38
rs771022969 Chr.11: 46719354-46719354 on Build GRCh38
rs958872946 Chr.11: 46719412-46719412 on Build GRCh38
rs3136431 Chr.11: 46719321-46719321 on Build GRCh38

View Full Product Details