Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42079650-42079911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMDT1
Entrez Gene ID
91689
Gene Name
single-pass membrane protein with aspartate rich tail 1
Gene Aliases
C22orf32, DDDD, EMRE, dJ186O1.1
UniGene
Hs.306083
Gene Chromosome Location
Chr.22: 42079687-42084284 on Build GRCh38
Transcripts
NM_033318.4 XM_011530509.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6559447 c.97G>T p.A33S Chr.22: 42079865-42079865 on Build GRCh38
3668613 c.87C>T p.G29G Chr.22: 42079855-42079855 on Build GRCh38
6785349 c.90T>C p.D30D Chr.22: 42079858-42079858 on Build GRCh38
6785350 c.129A>G p.V43V Chr.22: 42079897-42079897 on Build GRCh38
4667546 c.130C>T p.P44S Chr.22: 42079898-42079898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764866171 Chr.22: 42079892-42079892 on Build GRCh38
rs200780203 Chr.22: 42079788-42079788 on Build GRCh38
rs140698121 Chr.22: 42079838-42079838 on Build GRCh38
rs377070968 Chr.22: 42079831-42079831 on Build GRCh38
rs929680413 Chr.22: 42079737-42079737 on Build GRCh38
rs201691345 Chr.22: 42079793-42079793 on Build GRCh38
rs150077416 Chr.22: 42079856-42079856 on Build GRCh38
rs760008926 Chr.22: 42079881-42079881 on Build GRCh38
rs750717696 Chr.22: 42079721-42079721 on Build GRCh38
rs778939863 Chr.22: 42079748-42079748 on Build GRCh38
rs747295100 Chr.22: 42079767-42079767 on Build GRCh38
rs778689899 Chr.22: 42079827-42079827 on Build GRCh38
rs918343979 Chr.22: 42079715-42079715 on Build GRCh38
rs778836615 Chr.22: 42079843-42079843 on Build GRCh38
rs763841410 Chr.22: 42079808-42079808 on Build GRCh38
rs1023542162 Chr.22: 42079682-42079682 on Build GRCh38
rs946990414 Chr.22: 42079871-42079871 on Build GRCh38
rs999308566 Chr.22: 42079708-42079708 on Build GRCh38
rs2269524 Chr.22: 42079699-42079699 on Build GRCh38
rs982159427 Chr.22: 42079700-42079700 on Build GRCh38
rs758817205 Chr.22: 42079728-42079728 on Build GRCh38
rs773588630 Chr.22: 42079859-42079859 on Build GRCh38
rs755768479 Chr.22: 42079807-42079807 on Build GRCh38
rs755420668 Chr.22: 42079763-42079763 on Build GRCh38
rs766815239 Chr.22: 42079724-42079731 on Build GRCh38
rs752438036 Chr.22: 42079740-42079740 on Build GRCh38
rs144030548 Chr.22: 42079789-42079789 on Build GRCh38
rs907749381 Chr.22: 42079803-42079803 on Build GRCh38
rs765758120 Chr.22: 42079762-42079763 on Build GRCh38
rs545925762 Chr.22: 42079861-42079861 on Build GRCh38
rs757074571 Chr.22: 42079746-42079746 on Build GRCh38
rs746813471 Chr.22: 42079837-42079837 on Build GRCh38
rs767256057 Chr.22: 42079732-42079732 on Build GRCh38
rs752386703 Chr.22: 42079806-42079806 on Build GRCh38
rs768705774 Chr.22: 42079770-42079770 on Build GRCh38
rs775655465 Chr.22: 42079766-42079766 on Build GRCh38
rs763789816 Chr.22: 42079885-42079885 on Build GRCh38
rs376577255 Chr.22: 42079742-42079742 on Build GRCh38
rs376151257 Chr.22: 42079889-42079889 on Build GRCh38
rs774787944 Chr.22: 42079870-42079870 on Build GRCh38
rs574354973 Chr.22: 42079787-42079787 on Build GRCh38
rs1022493664 Chr.22: 42079689-42079689 on Build GRCh38
rs1046765 Chr.22: 42079878-42079878 on Build GRCh38
rs933956196 Chr.22: 42079840-42079840 on Build GRCh38
rs374562862 Chr.22: 42079850-42079850 on Build GRCh38
rs754411361 Chr.22: 42079751-42079752 on Build GRCh38
rs753245971 Chr.22: 42079820-42079820 on Build GRCh38
rs971009161 Chr.22: 42079868-42079868 on Build GRCh38
rs1049780318 Chr.22: 42079847-42079847 on Build GRCh38
rs763197879 Chr.22: 42079796-42079796 on Build GRCh38
rs776628036 Chr.22: 42079782-42079782 on Build GRCh38
rs766918669 Chr.22: 42079805-42079805 on Build GRCh38
rs369810620 Chr.22: 42079832-42079832 on Build GRCh38
rs745600257 Chr.22: 42079756-42079756 on Build GRCh38
rs748158819 Chr.22: 42079851-42079851 on Build GRCh38
rs753576232 Chr.22: 42079887-42079887 on Build GRCh38
rs150943700 Chr.22: 42079835-42079835 on Build GRCh38
rs756814054 Chr.22: 42079821-42079821 on Build GRCh38
rs990516039 Chr.22: 42079753-42079753 on Build GRCh38

View Full Product Details