Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127195288-127195545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27C1
Entrez Gene ID
339761
Gene Name
cytochrome P450 family 27 subfamily C member 1
Gene Aliases
-
UniGene
Hs.407639
Gene Chromosome Location
Chr.2: 127183832-127220096 on Build GRCh38
Transcripts
NM_001001665.3 XM_017003960.1 XM_017003961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5048920 c.695G>C p.R232T Chr.2: 127195359-127195359 on Build GRCh38
6087198 c.692T>C p.V231A Chr.2: 127195362-127195362 on Build GRCh38
4084666 c.629T>C p.I210T Chr.2: 127195425-127195425 on Build GRCh38
6466230 c.707A>T p.K236M Chr.2: 127195347-127195347 on Build GRCh38
4084665 c.650G>T p.R217M Chr.2: 127195404-127195404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779635468 Chr.2: 127195392-127195392 on Build GRCh38
rs147403081 Chr.2: 127195439-127195439 on Build GRCh38
rs756505793 Chr.2: 127195354-127195354 on Build GRCh38
rs754441587 Chr.2: 127195500-127195500 on Build GRCh38
rs769721923 Chr.2: 127195518-127195518 on Build GRCh38
rs776498930 Chr.2: 127195415-127195415 on Build GRCh38
rs569403160 Chr.2: 127195440-127195440 on Build GRCh38
rs749663934 Chr.2: 127195368-127195368 on Build GRCh38
rs751333445 Chr.2: 127195334-127195334 on Build GRCh38
rs752863092 Chr.2: 127195448-127195448 on Build GRCh38
rs199999961 Chr.2: 127195323-127195323 on Build GRCh38
rs7568070 Chr.2: 127195478-127195478 on Build GRCh38
rs868337701 Chr.2: 127195465-127195465 on Build GRCh38
rs564500900 Chr.2: 127195401-127195401 on Build GRCh38
rs533650259 Chr.2: 127195423-127195423 on Build GRCh38
rs768062477 Chr.2: 127195400-127195400 on Build GRCh38
rs530482051 Chr.2: 127195445-127195445 on Build GRCh38
rs746097771 Chr.2: 127195405-127195405 on Build GRCh38
rs147953391 Chr.2: 127195431-127195431 on Build GRCh38
rs758626622 Chr.2: 127195507-127195510 on Build GRCh38
rs780660849 Chr.2: 127195507-127195507 on Build GRCh38
rs763219322 Chr.2: 127195313-127195313 on Build GRCh38
rs147287859 Chr.2: 127195394-127195394 on Build GRCh38
rs373717507 Chr.2: 127195468-127195468 on Build GRCh38
rs771742150 Chr.2: 127195370-127195370 on Build GRCh38
rs767738253 Chr.2: 127195446-127195446 on Build GRCh38
rs779696383 Chr.2: 127195479-127195479 on Build GRCh38
rs150013191 Chr.2: 127195367-127195367 on Build GRCh38
rs766571172 Chr.2: 127195314-127195314 on Build GRCh38
rs756665338 Chr.2: 127195457-127195457 on Build GRCh38
rs746660693 Chr.2: 127195499-127195499 on Build GRCh38
rs142496522 Chr.2: 127195432-127195432 on Build GRCh38
rs747718320 Chr.2: 127195511-127195511 on Build GRCh38
rs1049583990 Chr.2: 127195497-127195497 on Build GRCh38
rs755017903 Chr.2: 127195327-127195327 on Build GRCh38
rs754225318 Chr.2: 127195473-127195473 on Build GRCh38
rs1036757398 Chr.2: 127195418-127195418 on Build GRCh38
rs748049896 Chr.2: 127195416-127195416 on Build GRCh38

View Full Product Details