Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173364900-173365133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
UniGene
Hs.470654
Gene Chromosome Location
Chr.2: 173354017-173368992 on Build GRCh38
Transcripts
NM_031942.4 NM_145810.2 XM_011511957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670751 c.834G>A p.E278E Chr.2: 173364929-173364929 on Build GRCh38
5860624 c.825_827delGGA p.E281delE Chr.2: 173364920-173364922 on Build GRCh38
6624521 c.866_867delAG p.R290fs*9 Chr.2: 173364961-173364962 on Build GRCh38
4086825 c.850T>C p.Y284H Chr.2: 173364945-173364945 on Build GRCh38
6427615 c.838G>T p.E280* Chr.2: 173364933-173364933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029961208 Chr.2: 173364919-173364919 on Build GRCh38
rs774465144 Chr.2: 173364975-173364975 on Build GRCh38
rs748445001 Chr.2: 173364945-173364945 on Build GRCh38
rs765456341 Chr.2: 173365001-173365001 on Build GRCh38
rs535908062 Chr.2: 173364922-173364922 on Build GRCh38
rs376689912 Chr.2: 173364982-173364982 on Build GRCh38
rs953939520 Chr.2: 173365026-173365026 on Build GRCh38
rs775230836 Chr.2: 173365041-173365041 on Build GRCh38
rs544697691 Chr.2: 173365098-173365098 on Build GRCh38
rs943883656 Chr.2: 173365080-173365080 on Build GRCh38
rs757432537 Chr.2: 173364919-173364920 on Build GRCh38
rs772146602 Chr.2: 173365067-173365067 on Build GRCh38
rs758958300 Chr.2: 173365033-173365033 on Build GRCh38
rs769997632 Chr.2: 173365025-173365025 on Build GRCh38
rs781543958 Chr.2: 173365017-173365017 on Build GRCh38
rs908943345 Chr.2: 173364953-173364953 on Build GRCh38
rs918645308 Chr.2: 173365093-173365093 on Build GRCh38
rs767491623 Chr.2: 173365036-173365036 on Build GRCh38
rs1000128877 Chr.2: 173364985-173364985 on Build GRCh38
rs779058449 Chr.2: 173364929-173364930 on Build GRCh38
rs780146971 Chr.2: 173365005-173365005 on Build GRCh38
rs893880946 Chr.2: 173365068-173365068 on Build GRCh38
rs55847722 Chr.2: 173365088-173365088 on Build GRCh38
rs1006964770 Chr.2: 173365076-173365076 on Build GRCh38
rs148421045 Chr.2: 173364971-173364971 on Build GRCh38
rs758309071 Chr.2: 173364918-173364918 on Build GRCh38
rs138353896 Chr.2: 173364976-173364976 on Build GRCh38
rs755565561 Chr.2: 173365013-173365013 on Build GRCh38
rs747068213 Chr.2: 173365010-173365010 on Build GRCh38
rs935349664 Chr.2: 173365040-173365040 on Build GRCh38
rs770680049 Chr.2: 173365030-173365030 on Build GRCh38
rs79281910 Chr.2: 173365028-173365028 on Build GRCh38
rs369071982 Chr.2: 173365000-173365000 on Build GRCh38
rs950446756 Chr.2: 173364942-173364942 on Build GRCh38
rs752598179 Chr.2: 173364997-173364997 on Build GRCh38
rs754798292 Chr.2: 173365053-173365053 on Build GRCh38
rs777100041 Chr.2: 173364940-173364940 on Build GRCh38
rs916458005 Chr.2: 173364989-173364989 on Build GRCh38
rs895361730 Chr.2: 173364944-173364944 on Build GRCh38
rs13382180 Chr.2: 173365003-173365003 on Build GRCh38
rs765939337 Chr.2: 173364972-173364972 on Build GRCh38
rs748552199 Chr.2: 173365023-173365023 on Build GRCh38
rs769558563 Chr.2: 173364964-173364964 on Build GRCh38
rs774057496 Chr.2: 173365031-173365031 on Build GRCh38
rs754083976 Chr.2: 173364920-173364922 on Build GRCh38

View Full Product Details