Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118339067-118339316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3D
Entrez Gene ID
915
Gene Name
CD3d molecule
Gene Aliases
CD3-DELTA, IMD19, T3D
UniGene
Hs.504048
Gene Chromosome Location
Chr.11: 118339074-118344233 on Build GRCh38
Transcripts
NM_000732.4 NM_001040651.1 XM_017018543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5679971 c.486C>A p.H162Q Chr.11: 118339192-118339192 on Build GRCh38
3868517 c.494G>A p.G165E Chr.11: 118339184-118339184 on Build GRCh38
274446 c.463C>T p.R155* Chr.11: 118339215-118339215 on Build GRCh38
923537 c.493G>T p.G165* Chr.11: 118339185-118339185 on Build GRCh38
3983804 c.505C>T p.R169W Chr.11: 118339173-118339173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745794915 Chr.11: 118339225-118339225 on Build GRCh38
rs375444192 Chr.11: 118339220-118339220 on Build GRCh38
rs777087947 Chr.11: 118339241-118339241 on Build GRCh38
rs758585177 Chr.11: 118339234-118339236 on Build GRCh38
rs775164531 Chr.11: 118339117-118339117 on Build GRCh38
rs768234684 Chr.11: 118339236-118339236 on Build GRCh38
rs567299359 Chr.11: 118339201-118339201 on Build GRCh38
rs753689641 Chr.11: 118339258-118339258 on Build GRCh38
rs751892269 Chr.11: 118339202-118339202 on Build GRCh38
rs201045543 Chr.11: 118339175-118339175 on Build GRCh38
rs771349801 Chr.11: 118339121-118339121 on Build GRCh38
rs201691963 Chr.11: 118339264-118339264 on Build GRCh38
rs975995932 Chr.11: 118339207-118339207 on Build GRCh38
rs113271462 Chr.11: 118339103-118339103 on Build GRCh38
rs368739909 Chr.11: 118339267-118339267 on Build GRCh38
rs367800432 Chr.11: 118339173-118339173 on Build GRCh38
rs760334947 Chr.11: 118339230-118339230 on Build GRCh38
rs771195985 Chr.11: 118339213-118339213 on Build GRCh38
rs764502874 Chr.11: 118339133-118339133 on Build GRCh38
rs2276424 Chr.11: 118339245-118339245 on Build GRCh38
rs750498030 Chr.11: 118339226-118339227 on Build GRCh38
rs201994476 Chr.11: 118339221-118339221 on Build GRCh38
rs571561124 Chr.11: 118339090-118339090 on Build GRCh38
rs868824577 Chr.11: 118339225-118339226 on Build GRCh38
rs374400896 Chr.11: 118339283-118339283 on Build GRCh38
rs146997233 Chr.11: 118339168-118339168 on Build GRCh38
rs200731099 Chr.11: 118339137-118339137 on Build GRCh38
rs750542171 Chr.11: 118339170-118339170 on Build GRCh38
rs757790037 Chr.11: 118339163-118339163 on Build GRCh38
rs771637289 Chr.11: 118339227-118339227 on Build GRCh38
rs763545091 Chr.11: 118339251-118339251 on Build GRCh38
rs201961490 Chr.11: 118339249-118339249 on Build GRCh38
rs760316469 Chr.11: 118339122-118339122 on Build GRCh38
rs200108243 Chr.11: 118339172-118339172 on Build GRCh38
rs777721098 Chr.11: 118339215-118339215 on Build GRCh38
rs755253686 Chr.11: 118339253-118339253 on Build GRCh38
rs200942033 Chr.11: 118339250-118339250 on Build GRCh38
rs974588593 Chr.11: 118339136-118339136 on Build GRCh38
rs201271197 Chr.11: 118339252-118339252 on Build GRCh38
rs781522024 Chr.11: 118339254-118339254 on Build GRCh38
rs538224426 Chr.11: 118339280-118339280 on Build GRCh38
rs755235102 Chr.11: 118339206-118339206 on Build GRCh38

View Full Product Details