Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57801961-57802230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND1
Entrez Gene ID
1500
Gene Name
catenin delta 1
Gene Aliases
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57761762-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654668 c.1381C>T p.R461* Chr.11: 57802157-57802157 on Build GRCh38
928712 c.1241T>C p.V414A Chr.11: 57802017-57802017 on Build GRCh38
3449776 c.1402C>A p.L468I Chr.11: 57802178-57802178 on Build GRCh38
928714 c.1342A>C p.K448Q Chr.11: 57802118-57802118 on Build GRCh38
6133605 c.1190A>G p.N397S Chr.11: 57801966-57801966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777580736 Chr.11: 57802184-57802184 on Build GRCh38
rs759069892 Chr.11: 57802051-57802051 on Build GRCh38
rs760079351 Chr.11: 57802009-57802009 on Build GRCh38
rs780597185 Chr.11: 57802139-57802139 on Build GRCh38
rs770867646 Chr.11: 57802049-57802049 on Build GRCh38
rs200755967 Chr.11: 57801982-57801982 on Build GRCh38
rs759322791 Chr.11: 57802202-57802202 on Build GRCh38
rs767138391 Chr.11: 57802068-57802068 on Build GRCh38
rs769966903 Chr.11: 57802181-57802181 on Build GRCh38
rs199779076 Chr.11: 57802195-57802195 on Build GRCh38
rs755094282 Chr.11: 57802164-57802164 on Build GRCh38
rs760344044 Chr.11: 57802082-57802082 on Build GRCh38
rs1031132836 Chr.11: 57802153-57802153 on Build GRCh38
rs1026452009 Chr.11: 57802022-57802022 on Build GRCh38
rs752398112 Chr.11: 57802070-57802070 on Build GRCh38
rs751657184 Chr.11: 57802158-57802158 on Build GRCh38
rs759024257 Chr.11: 57802092-57802092 on Build GRCh38
rs770234783 Chr.11: 57802132-57802132 on Build GRCh38
rs541781874 Chr.11: 57801985-57801985 on Build GRCh38
rs537846966 Chr.11: 57802087-57802087 on Build GRCh38
rs775746543 Chr.11: 57802146-57802146 on Build GRCh38
rs763306444 Chr.11: 57802167-57802167 on Build GRCh38
rs762912441 Chr.11: 57802045-57802045 on Build GRCh38
rs771623674 Chr.11: 57802204-57802204 on Build GRCh38
rs371034991 Chr.11: 57802091-57802091 on Build GRCh38
rs749055749 Chr.11: 57802186-57802186 on Build GRCh38
rs11570199 Chr.11: 57802166-57802166 on Build GRCh38

View Full Product Details