Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49097319-49097590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYM
Entrez Gene ID
54808
Gene Name
dymeclin
Gene Aliases
DMC, SMC
UniGene
Hs.162996
Gene Chromosome Location
Chr.18: 49041474-49460709 on Build GRCh38
Transcripts
NM_017653.3 XM_006722488.2 XM_006722490.2 XM_011526036.1 XM_011526037.1 XM_011526038.1 XM_011526039.1 XM_011526040.1 XM_011526041.1 XM_011526042.1 XM_017025795.1 XM_017025796.1 XM_017025797.1 XM_017025798.1 XM_017025799.1 XM_017025800.1 XM_017025801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5141195 c.1860+5G>T p.? Chr.18: 49097397-49097397 on Build GRCh38
1204751 c.1805G>A p.R602Q Chr.18: 49097457-49097457 on Build GRCh38
4532024 c.1849G>A p.D617N Chr.18: 49097413-49097413 on Build GRCh38
3362537 c.1860G>T p.K620N Chr.18: 49097402-49097402 on Build GRCh38
3734362 c.1760T>C p.F587S Chr.18: 49097502-49097502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767368342 Chr.18: 49097365-49097365 on Build GRCh38
rs747450845 Chr.18: 49097399-49097399 on Build GRCh38
rs373233127 Chr.18: 49097366-49097366 on Build GRCh38
rs972757417 Chr.18: 49097545-49097545 on Build GRCh38
rs151034190 Chr.18: 49097503-49097503 on Build GRCh38
rs764087116 Chr.18: 49097530-49097530 on Build GRCh38
rs146000214 Chr.18: 49097484-49097484 on Build GRCh38
rs771839172 Chr.18: 49097492-49097492 on Build GRCh38
rs786205511 Chr.18: 49097401-49097401 on Build GRCh38
rs761513723 Chr.18: 49097431-49097431 on Build GRCh38
rs756046941 Chr.18: 49097372-49097372 on Build GRCh38
rs1053113624 Chr.18: 49097552-49097552 on Build GRCh38
rs774241781 Chr.18: 49097357-49097357 on Build GRCh38
rs768403608 Chr.18: 49097428-49097428 on Build GRCh38
rs773080872 Chr.18: 49097433-49097433 on Build GRCh38
rs1006114843 Chr.18: 49097499-49097499 on Build GRCh38
rs773893296 Chr.18: 49097429-49097429 on Build GRCh38
rs377444815 Chr.18: 49097466-49097466 on Build GRCh38
rs760754978 Chr.18: 49097437-49097437 on Build GRCh38
rs781551662 Chr.18: 49097458-49097458 on Build GRCh38
rs376308649 Chr.18: 49097491-49097491 on Build GRCh38
rs748714932 Chr.18: 49097425-49097425 on Build GRCh38
rs141363417 Chr.18: 49097505-49097505 on Build GRCh38
rs768078815 Chr.18: 49097474-49097474 on Build GRCh38
rs777830697 Chr.18: 49097393-49097393 on Build GRCh38
rs371247672 Chr.18: 49097475-49097475 on Build GRCh38
rs368200347 Chr.18: 49097500-49097500 on Build GRCh38
rs60818038 Chr.18: 49097513-49097513 on Build GRCh38
rs757405720 Chr.18: 49097453-49097453 on Build GRCh38
rs991362073 Chr.18: 49097465-49097465 on Build GRCh38
rs1011113052 Chr.18: 49097380-49097380 on Build GRCh38
rs746380133 Chr.18: 49097412-49097412 on Build GRCh38
rs751582588 Chr.18: 49097562-49097562 on Build GRCh38
rs201023000 Chr.18: 49097450-49097450 on Build GRCh38
rs770269902 Chr.18: 49097423-49097423 on Build GRCh38
rs374799304 Chr.18: 49097452-49097452 on Build GRCh38
rs765423036 Chr.18: 49097384-49097384 on Build GRCh38
rs776455503 Chr.18: 49097510-49097510 on Build GRCh38
rs370290857 Chr.18: 49097432-49097432 on Build GRCh38
rs745978580 Chr.18: 49097444-49097444 on Build GRCh38
rs757641481 Chr.18: 49097407-49097407 on Build GRCh38
rs752865908 Chr.18: 49097446-49097446 on Build GRCh38
rs750780184 Chr.18: 49097464-49097464 on Build GRCh38
rs763432291 Chr.18: 49097534-49097534 on Build GRCh38
rs201477312 Chr.18: 49097424-49097424 on Build GRCh38
rs142608802 Chr.18: 49097410-49097410 on Build GRCh38
rs752710018 Chr.18: 49097389-49097389 on Build GRCh38
rs761677430 Chr.18: 49097359-49097359 on Build GRCh38

View Full Product Details