Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12477439-12477668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP16
Entrez Gene ID
80824
Gene Name
dual specificity phosphatase 16
Gene Aliases
MKP-7, MKP7
UniGene
Hs.536535
Gene Chromosome Location
Chr.12: 12473282-12562839 on Build GRCh38
Transcripts
NM_030640.2 XM_005253488.2 XM_006719155.2 XM_011520856.1 XM_011520857.1 XM_017019988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2227589 c.1252C>T p.H418Y Chr.12: 12477579-12477579 on Build GRCh38
6519979 c.1356G>T p.S452S Chr.12: 12477475-12477475 on Build GRCh38
5027542 c.1204C>G p.L402V Chr.12: 12477627-12477627 on Build GRCh38
430689 c.1287C>G p.Y429* Chr.12: 12477544-12477544 on Build GRCh38
4679709 c.1390G>T p.E464* Chr.12: 12477441-12477441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760515668 Chr.12: 12477595-12477595 on Build GRCh38
rs759667155 Chr.12: 12477491-12477491 on Build GRCh38
rs143980194 Chr.12: 12477628-12477628 on Build GRCh38
rs751940406 Chr.12: 12477524-12477524 on Build GRCh38
rs199972119 Chr.12: 12477523-12477523 on Build GRCh38
rs200519285 Chr.12: 12477489-12477489 on Build GRCh38
rs759509351 Chr.12: 12477606-12477606 on Build GRCh38
rs907275967 Chr.12: 12477527-12477527 on Build GRCh38
rs374358053 Chr.12: 12477620-12477620 on Build GRCh38
rs771041816 Chr.12: 12477598-12477598 on Build GRCh38
rs754592876 Chr.12: 12477593-12477593 on Build GRCh38
rs767817494 Chr.12: 12477638-12477638 on Build GRCh38
rs372494052 Chr.12: 12477565-12477565 on Build GRCh38
rs373426579 Chr.12: 12477496-12477496 on Build GRCh38
rs775580594 Chr.12: 12477609-12477609 on Build GRCh38
rs536844023 Chr.12: 12477579-12477579 on Build GRCh38
rs748787807 Chr.12: 12477462-12477462 on Build GRCh38
rs149924901 Chr.12: 12477569-12477569 on Build GRCh38
rs747771857 Chr.12: 12477577-12477577 on Build GRCh38
rs765234439 Chr.12: 12477608-12477608 on Build GRCh38
rs756589989 Chr.12: 12477646-12477646 on Build GRCh38
rs776733348 Chr.12: 12477600-12477600 on Build GRCh38
rs763142603 Chr.12: 12477612-12477612 on Build GRCh38
rs773850223 Chr.12: 12477470-12477470 on Build GRCh38
rs752892100 Chr.12: 12477510-12477510 on Build GRCh38
rs568189181 Chr.12: 12477530-12477530 on Build GRCh38
rs757592289 Chr.12: 12477634-12477634 on Build GRCh38
rs371354932 Chr.12: 12477525-12477525 on Build GRCh38
rs139164615 Chr.12: 12477472-12477472 on Build GRCh38
rs369438527 Chr.12: 12477475-12477475 on Build GRCh38
rs768334806 Chr.12: 12477469-12477469 on Build GRCh38
rs201932444 Chr.12: 12477542-12477542 on Build GRCh38
rs527241198 Chr.12: 12477639-12477639 on Build GRCh38
rs758578113 Chr.12: 12477511-12477511 on Build GRCh38
rs778422571 Chr.12: 12477558-12477558 on Build GRCh38
rs768175453 Chr.12: 12477552-12477552 on Build GRCh38
rs748842953 Chr.12: 12477547-12477547 on Build GRCh38

View Full Product Details