Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50310069-50310336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
UniGene
Hs.130310
Gene Chromosome Location
Chr.X: 50202713-50351914 on Build GRCh38
Transcripts
NM_033031.2 NM_033670.2 XM_017029913.1 XM_017029914.1 XM_017029915.1 XM_017029916.1 XM_017029917.1 XM_017029918.1

SNP Detail

SNP ID Chromosome Location
rs370387578 Chr.X: 50310101-50310101 on Build GRCh38
rs377732336 Chr.X: 50310152-50310152 on Build GRCh38
rs201598054 Chr.X: 50310116-50310116 on Build GRCh38
rs782385072 Chr.X: 50310245-50310245 on Build GRCh38
rs781818274 Chr.X: 50310148-50310148 on Build GRCh38
rs782162812 Chr.X: 50310104-50310104 on Build GRCh38
rs375731852 Chr.X: 50310306-50310306 on Build GRCh38
rs372453177 Chr.X: 50310150-50310150 on Build GRCh38
rs782423834 Chr.X: 50310112-50310112 on Build GRCh38
rs782249730 Chr.X: 50310217-50310217 on Build GRCh38
rs782607954 Chr.X: 50310091-50310091 on Build GRCh38
rs782756366 Chr.X: 50310151-50310151 on Build GRCh38
rs782776724 Chr.X: 50310222-50310222 on Build GRCh38
rs73213610 Chr.X: 50310275-50310275 on Build GRCh38
rs782309824 Chr.X: 50310233-50310233 on Build GRCh38
rs782483360 Chr.X: 50310191-50310191 on Build GRCh38
rs139254834 Chr.X: 50310254-50310254 on Build GRCh38
rs781939355 Chr.X: 50310119-50310119 on Build GRCh38
rs889888532 Chr.X: 50310144-50310144 on Build GRCh38
rs782313834 Chr.X: 50310227-50310227 on Build GRCh38
rs994453050 Chr.X: 50310234-50310234 on Build GRCh38
rs782652552 Chr.X: 50310207-50310207 on Build GRCh38
rs782524197 Chr.X: 50310180-50310180 on Build GRCh38
rs192785609 Chr.X: 50310276-50310276 on Build GRCh38
rs782202179 Chr.X: 50310220-50310220 on Build GRCh38
rs781787863 Chr.X: 50310182-50310182 on Build GRCh38
rs267606474 Chr.X: 50310127-50310127 on Build GRCh38
rs1051215074 Chr.X: 50310113-50310113 on Build GRCh38
rs782200903 Chr.X: 50310237-50310237 on Build GRCh38
rs782673169 Chr.X: 50310221-50310221 on Build GRCh38
rs782051243 Chr.X: 50310118-50310119 on Build GRCh38
rs1017358207 Chr.X: 50310153-50310153 on Build GRCh38
rs782198463 Chr.X: 50310225-50310225 on Build GRCh38
rs146219939 Chr.X: 50310134-50310134 on Build GRCh38
rs782057635 Chr.X: 50310128-50310128 on Build GRCh38
rs782014132 Chr.X: 50310103-50310103 on Build GRCh38

View Full Product Details