Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29793404-29793643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
UniGene
Hs.731754
Gene Chromosome Location
Chr.22: 29788608-29838304 on Build GRCh38
Transcripts
NM_001242906.1 NM_032204.4 XM_005261775.2 XM_011530442.2 XM_011530443.2 XM_011530444.2 XM_011530445.2 XM_011530446.2 XM_011530448.2 XM_011530449.2 XM_011530450.2 XM_011530451.2 XM_011530452.2 XM_011530453.2 XM_011530454.2 XM_011530455.2 XM_017028991.1 XM_017028992.1 XM_017028993.1 XM_017028994.1 XM_017028995.1 XM_017028996.1 XM_017028997.1 XM_017028998.1 XM_017028999.1 XM_017029000.1 XM_017029001.1 XM_017029002.1 XM_017029003.1 XM_017029004.1 XM_017029005.1 XM_017029006.1 XM_017029007.1 XM_017029008.1 XM_017029009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553610 c.1784A>T p.E595V Chr.22: 29793581-29793581 on Build GRCh38
1495188 c.1792C>A p.P598T Chr.22: 29793487-29793487 on Build GRCh38
4968670 c.1732C>T p.R578C Chr.22: 29793633-29793633 on Build GRCh38
444857 c.1860C>T p.Y620Y Chr.22: 29793419-29793419 on Build GRCh38
725979 c.1809G>C p.E603D Chr.22: 29793470-29793470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143291076 Chr.22: 29793608-29793608 on Build GRCh38
rs200305556 Chr.22: 29793494-29793494 on Build GRCh38
rs765944174 Chr.22: 29793624-29793624 on Build GRCh38
rs759695416 Chr.22: 29793478-29793478 on Build GRCh38
rs551797035 Chr.22: 29793614-29793614 on Build GRCh38
rs1001583779 Chr.22: 29793525-29793525 on Build GRCh38
rs767825721 Chr.22: 29793621-29793621 on Build GRCh38
rs747401908 Chr.22: 29793558-29793558 on Build GRCh38
rs367805592 Chr.22: 29793523-29793523 on Build GRCh38
rs747990302 Chr.22: 29793564-29793564 on Build GRCh38
rs908439553 Chr.22: 29793474-29793474 on Build GRCh38
rs777026011 Chr.22: 29793583-29793583 on Build GRCh38
rs529377081 Chr.22: 29793514-29793514 on Build GRCh38
rs768389612 Chr.22: 29793443-29793443 on Build GRCh38
rs368266999 Chr.22: 29793453-29793453 on Build GRCh38
rs763494159 Chr.22: 29793498-29793498 on Build GRCh38
rs760876798 Chr.22: 29793467-29793467 on Build GRCh38
rs774168551 Chr.22: 29793461-29793461 on Build GRCh38
rs775679720 Chr.22: 29793591-29793591 on Build GRCh38
rs146488197 Chr.22: 29793472-29793472 on Build GRCh38
rs750094453 Chr.22: 29793622-29793622 on Build GRCh38
rs755648404 Chr.22: 29793518-29793518 on Build GRCh38
rs896015672 Chr.22: 29793516-29793516 on Build GRCh38
rs768636755 Chr.22: 29793528-29793528 on Build GRCh38
rs763435583 Chr.22: 29793592-29793592 on Build GRCh38
rs773513225 Chr.22: 29793478-29793479 on Build GRCh38
rs372907029 Chr.22: 29793505-29793505 on Build GRCh38
rs779967582 Chr.22: 29793440-29793440 on Build GRCh38
rs759784246 Chr.22: 29793586-29793586 on Build GRCh38
rs749227680 Chr.22: 29793526-29793526 on Build GRCh38
rs780631424 Chr.22: 29793424-29793424 on Build GRCh38
rs769232988 Chr.22: 29793435-29793435 on Build GRCh38
rs34062345 Chr.22: 29793603-29793603 on Build GRCh38
rs776938110 Chr.22: 29793473-29793473 on Build GRCh38
rs745367051 Chr.22: 29793431-29793431 on Build GRCh38
rs1055767039 Chr.22: 29793531-29793531 on Build GRCh38
rs185631950 Chr.22: 29793515-29793515 on Build GRCh38
rs770043157 Chr.22: 29793589-29793589 on Build GRCh38
rs569408700 Chr.22: 29793567-29793567 on Build GRCh38
rs112106561 Chr.22: 29793508-29793508 on Build GRCh38
rs964205248 Chr.22: 29793609-29793609 on Build GRCh38
rs371701937 Chr.22: 29793550-29793550 on Build GRCh38
rs34564532 Chr.22: 29793623-29793623 on Build GRCh38
rs749179340 Chr.22: 29793442-29793442 on Build GRCh38

View Full Product Details