Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10556772-10556997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT2
Entrez Gene ID
2651
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
UniGene
Hs.519884
Gene Chromosome Location
Chr.6: 10521283-10629368 on Build GRCh38
Transcripts
NM_001491.2 NM_145649.4 XM_005248997.3 XM_005248999.2 XM_006715052.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1071712 c.376G>A p.V126M Chr.6: 10556799-10556799 on Build GRCh38
3366242 c.367T>G p.C123G Chr.6: 10556790-10556790 on Build GRCh38
4160090 c.459G>T p.K153N Chr.6: 10556882-10556882 on Build GRCh38
5522748 c.523C>T p.L175F Chr.6: 10556946-10556946 on Build GRCh38
4498979 c.531C>T p.A177A Chr.6: 10556954-10556954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551675854 Chr.6: 10556962-10556962 on Build GRCh38
rs202207478 Chr.6: 10556906-10556906 on Build GRCh38
rs147742441 Chr.6: 10556946-10556946 on Build GRCh38
rs535689181 Chr.6: 10556911-10556911 on Build GRCh38
rs113836537 Chr.6: 10556817-10556817 on Build GRCh38
rs147047890 Chr.6: 10556899-10556899 on Build GRCh38
rs1024307909 Chr.6: 10556832-10556832 on Build GRCh38
rs762675289 Chr.6: 10556923-10556924 on Build GRCh38
rs200102997 Chr.6: 10556935-10556935 on Build GRCh38
rs778682112 Chr.6: 10556908-10556908 on Build GRCh38
rs560637463 Chr.6: 10556940-10556940 on Build GRCh38
rs758306131 Chr.6: 10556918-10556918 on Build GRCh38
rs754744022 Chr.6: 10556802-10556802 on Build GRCh38
rs142662339 Chr.6: 10556950-10556950 on Build GRCh38
rs777810837 Chr.6: 10556927-10556927 on Build GRCh38
rs768135788 Chr.6: 10556972-10556972 on Build GRCh38
rs753676781 Chr.6: 10556843-10556843 on Build GRCh38
rs764700830 Chr.6: 10556847-10556848 on Build GRCh38
rs148669958 Chr.6: 10556808-10556808 on Build GRCh38
rs147905083 Chr.6: 10556891-10556891 on Build GRCh38
rs753463085 Chr.6: 10556884-10556884 on Build GRCh38
rs769030276 Chr.6: 10556963-10556963 on Build GRCh38
rs770979858 Chr.6: 10556937-10556937 on Build GRCh38
rs539812897 Chr.6: 10556833-10556833 on Build GRCh38
rs770940749 Chr.6: 10556826-10556826 on Build GRCh38
rs1027713932 Chr.6: 10556857-10556857 on Build GRCh38
rs142308624 Chr.6: 10556865-10556865 on Build GRCh38
rs1001635904 Chr.6: 10556851-10556851 on Build GRCh38
rs141492136 Chr.6: 10556892-10556892 on Build GRCh38
rs199770755 Chr.6: 10556975-10556975 on Build GRCh38
rs774943999 Chr.6: 10556910-10556910 on Build GRCh38
rs747051358 Chr.6: 10556819-10556819 on Build GRCh38
rs763265359 Chr.6: 10556958-10556958 on Build GRCh38
rs774780221 Chr.6: 10556967-10556967 on Build GRCh38
rs764556134 Chr.6: 10556864-10556864 on Build GRCh38
rs748020006 Chr.6: 10556811-10556811 on Build GRCh38
rs960251969 Chr.6: 10556895-10556895 on Build GRCh38
rs201167779 Chr.6: 10556855-10556855 on Build GRCh38
rs903709186 Chr.6: 10556815-10556815 on Build GRCh38
rs762230655 Chr.6: 10556971-10556971 on Build GRCh38
rs763357071 Chr.6: 10556860-10556860 on Build GRCh38
rs764763524 Chr.6: 10556976-10556976 on Build GRCh38
rs765818421 Chr.6: 10556871-10556871 on Build GRCh38
rs760128376 Chr.6: 10556868-10556868 on Build GRCh38
rs896391023 Chr.6: 10556821-10556821 on Build GRCh38
rs373471539 Chr.6: 10556800-10556800 on Build GRCh38
rs970425612 Chr.6: 10556846-10556846 on Build GRCh38
rs746019094 Chr.6: 10556943-10556943 on Build GRCh38

View Full Product Details