Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127052514-127052739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
UniGene
Hs.49573
Gene Chromosome Location
Chr.5: 127040752-127073539 on Build GRCh38
Transcripts
NM_001164478.1 NM_001164479.1 XM_017009464.1 XM_017009465.1

SNP Detail

SNP ID Chromosome Location
rs745777421 Chr.5: 127052659-127052659 on Build GRCh38
rs775226437 Chr.5: 127052637-127052637 on Build GRCh38
rs374041179 Chr.5: 127052658-127052658 on Build GRCh38
rs1019713437 Chr.5: 127052643-127052643 on Build GRCh38
rs111426882 Chr.5: 127052698-127052698 on Build GRCh38
rs144574032 Chr.5: 127052671-127052671 on Build GRCh38
rs1050714516 Chr.5: 127052622-127052622 on Build GRCh38
rs769377129 Chr.5: 127052620-127052620 on Build GRCh38
rs542774691 Chr.5: 127052713-127052713 on Build GRCh38
rs140808546 Chr.5: 127052609-127052609 on Build GRCh38
rs537950236 Chr.5: 127052610-127052610 on Build GRCh38
rs745500328 Chr.5: 127052592-127052592 on Build GRCh38
rs1000377016 Chr.5: 127052695-127052695 on Build GRCh38
rs777748493 Chr.5: 127052618-127052618 on Build GRCh38
rs753609698 Chr.5: 127052679-127052680 on Build GRCh38
rs921194752 Chr.5: 127052564-127052564 on Build GRCh38
rs780631317 Chr.5: 127052588-127052588 on Build GRCh38
rs1043867007 Chr.5: 127052681-127052681 on Build GRCh38
rs370278047 Chr.5: 127052694-127052694 on Build GRCh38

View Full Product Details