Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186786163-186786436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A2
Entrez Gene ID
1974
Gene Name
eukaryotic translation initiation factor 4A2
Gene Aliases
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186783572-186789897 on Build GRCh38
Transcripts
NM_001967.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6468737 c.603C>T p.F201F Chr.3: 186786249-186786249 on Build GRCh38
3846966 c.595G>A p.E199K Chr.3: 186786241-186786241 on Build GRCh38
7112317 c.536_539delTGTT p.V181fs*8 Chr.3: 186786182-186786185 on Build GRCh38
446155 c.518-1G>A p.? Chr.3: 186786163-186786163 on Build GRCh38
446156 c.595G>C p.E199Q Chr.3: 186786241-186786241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768409704 Chr.3: 186786212-186786213 on Build GRCh38
rs763410755 Chr.3: 186786280-186786280 on Build GRCh38
rs981943557 Chr.3: 186786340-186786340 on Build GRCh38
rs1035927651 Chr.3: 186786323-186786323 on Build GRCh38
rs755015035 Chr.3: 186786324-186786324 on Build GRCh38
rs73886079 Chr.3: 186786386-186786386 on Build GRCh38
rs369787646 Chr.3: 186786390-186786390 on Build GRCh38
rs901737107 Chr.3: 186786405-186786405 on Build GRCh38
rs750878467 Chr.3: 186786314-186786314 on Build GRCh38
rs550464771 Chr.3: 186786330-186786330 on Build GRCh38
rs746312331 Chr.3: 186786412-186786426 on Build GRCh38
rs533856697 Chr.3: 186786375-186786375 on Build GRCh38
rs747792520 Chr.3: 186786302-186786302 on Build GRCh38
rs1038956161 Chr.3: 186786395-186786395 on Build GRCh38
rs748463432 Chr.3: 186786288-186786288 on Build GRCh38
rs266718 Chr.3: 186786359-186786359 on Build GRCh38
rs890647031 Chr.3: 186786394-186786394 on Build GRCh38
rs759291333 Chr.3: 186786294-186786294 on Build GRCh38
rs755882910 Chr.3: 186786231-186786231 on Build GRCh38
rs770152296 Chr.3: 186786289-186786289 on Build GRCh38
rs886413699 Chr.3: 186786361-186786363 on Build GRCh38
rs539100607 Chr.3: 186786352-186786352 on Build GRCh38
rs369188077 Chr.3: 186786318-186786318 on Build GRCh38
rs377484654 Chr.3: 186786312-186786312 on Build GRCh38
rs765913338 Chr.3: 186786293-186786293 on Build GRCh38
rs200193389 Chr.3: 186786296-186786296 on Build GRCh38
rs77264282 Chr.3: 186786350-186786350 on Build GRCh38
rs979582770 Chr.3: 186786370-186786370 on Build GRCh38
rs771316701 Chr.3: 186786288-186786289 on Build GRCh38
rs34071953 Chr.3: 186786371-186786372 on Build GRCh38
rs370965551 Chr.3: 186786280-186786280 on Build GRCh38
rs747278362 Chr.3: 186786283-186786283 on Build GRCh38
rs373878541 Chr.3: 186786305-186786305 on Build GRCh38
rs369523658 Chr.3: 186786292-186786292 on Build GRCh38
rs1047513519 Chr.3: 186786412-186786412 on Build GRCh38
rs960224456 Chr.3: 186786338-186786338 on Build GRCh38
rs933170461 Chr.3: 186786408-186786408 on Build GRCh38
rs529026789 Chr.3: 186786310-186786310 on Build GRCh38
rs773441667 Chr.3: 186786265-186786266 on Build GRCh38
rs761040011 Chr.3: 186786327-186786327 on Build GRCh38
rs564616540 Chr.3: 186786395-186786396 on Build GRCh38
rs575110411 Chr.3: 186786393-186786393 on Build GRCh38
rs62294447 Chr.3: 186786414-186786414 on Build GRCh38
rs375367664 Chr.3: 186786282-186786282 on Build GRCh38
rs749201932 Chr.3: 186786266-186786266 on Build GRCh38
rs777858280 Chr.3: 186786234-186786234 on Build GRCh38
rs886531983 Chr.3: 186786413-186786413 on Build GRCh38
rs546420930 Chr.3: 186786255-186786255 on Build GRCh38
rs781298566 Chr.3: 186786325-186786325 on Build GRCh38
rs763935517 Chr.3: 186786303-186786303 on Build GRCh38
rs776850844 Chr.3: 186786284-186786284 on Build GRCh38
rs774864262 Chr.3: 186786324-186786325 on Build GRCh38
rs566473858 Chr.3: 186786360-186786360 on Build GRCh38

View Full Product Details