Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67831521-67831770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4261304 c.198G>T p.G66G Chr.16: 67831770-67831770 on Build GRCh38
4261303 c.214G>T p.E72* Chr.16: 67831754-67831754 on Build GRCh38
5983804 c.220C>A p.Q74K Chr.16: 67831607-67831607 on Build GRCh38
1679191 c.523G>T p.E175* Chr.16: 67831754-67831754 on Build GRCh38
5983803 c.529C>A p.Q177K Chr.16: 67831607-67831607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771328291 Chr.16: 67831728-67831728 on Build GRCh38
rs962513990 Chr.16: 67831605-67831605 on Build GRCh38
rs185855516 Chr.16: 67831704-67831704 on Build GRCh38
rs558638104 Chr.16: 67831679-67831679 on Build GRCh38
rs557075852 Chr.16: 67831653-67831653 on Build GRCh38
rs760885247 Chr.16: 67831724-67831724 on Build GRCh38
rs753297172 Chr.16: 67831632-67831633 on Build GRCh38
rs371104081 Chr.16: 67831563-67831563 on Build GRCh38
rs376945315 Chr.16: 67831541-67831541 on Build GRCh38
rs566649456 Chr.16: 67831625-67831625 on Build GRCh38
rs997486445 Chr.16: 67831544-67831544 on Build GRCh38
rs534238356 Chr.16: 67831678-67831678 on Build GRCh38
rs1030765295 Chr.16: 67831557-67831557 on Build GRCh38
rs8056617 Chr.16: 67831596-67831596 on Build GRCh38
rs778553356 Chr.16: 67831713-67831713 on Build GRCh38
rs781123654 Chr.16: 67831583-67831583 on Build GRCh38
rs768028004 Chr.16: 67831659-67831659 on Build GRCh38
rs570946463 Chr.16: 67831698-67831698 on Build GRCh38
rs144356336 Chr.16: 67831718-67831718 on Build GRCh38
rs750254179 Chr.16: 67831616-67831616 on Build GRCh38
rs776816049 Chr.16: 67831732-67831732 on Build GRCh38
rs370602284 Chr.16: 67831613-67831613 on Build GRCh38
rs375155332 Chr.16: 67831719-67831719 on Build GRCh38
rs765224013 Chr.16: 67831745-67831745 on Build GRCh38
rs773571241 Chr.16: 67831552-67831552 on Build GRCh38
rs751442028 Chr.16: 67831599-67831599 on Build GRCh38
rs748536924 Chr.16: 67831715-67831715 on Build GRCh38
rs903753802 Chr.16: 67831624-67831624 on Build GRCh38
rs761062377 Chr.16: 67831726-67831726 on Build GRCh38
rs928278410 Chr.16: 67831735-67831739 on Build GRCh38
rs779882026 Chr.16: 67831628-67831628 on Build GRCh38
rs983484970 Chr.16: 67831570-67831570 on Build GRCh38
rs760831431 Chr.16: 67831554-67831554 on Build GRCh38
rs1045378076 Chr.16: 67831641-67831642 on Build GRCh38
rs759863422 Chr.16: 67831740-67831740 on Build GRCh38
rs757282392 Chr.16: 67831608-67831608 on Build GRCh38
rs948665707 Chr.16: 67831635-67831635 on Build GRCh38
rs748988901 Chr.16: 67831641-67831641 on Build GRCh38

View Full Product Details