Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45117683-45117918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOXA1
Entrez Gene ID
90527
Gene Name
dual oxidase maturation factor 1
Gene Aliases
NIP, NUMBIP, mol
UniGene
Hs.356664
Gene Chromosome Location
Chr.15: 45117366-45129938 on Build GRCh38
Transcripts
NM_001276264.1 NM_001276265.1 NM_001276267.1 NM_001276268.1 NM_144565.3 XM_006720743.3 XM_006720745.1 XM_006720746.1 XM_006720747.3 XM_006720749.1 XM_006720751.3 XM_006720752.3 XM_006720754.1 XM_006720755.1 XM_011522181.2 XM_011522182.1 XM_011522183.2 NM_207581.3 XM_017022180.1

Gene Symbol
DUOXA2
Entrez Gene ID
405753
Gene Name
dual oxidase maturation factor 2
Gene Aliases
SIMNIPHOM, TDH5
UniGene
Hs.497987
Gene Chromosome Location
Chr.15: 45114321-45118106 on Build GRCh38
Transcripts
NM_001276264.1 NM_001276265.1 NM_001276267.1 NM_001276268.1 NM_144565.3 XM_006720743.3 XM_006720745.1 XM_006720746.1 XM_006720747.3 XM_006720749.1 XM_006720751.3 XM_006720752.3 XM_006720754.1 XM_006720755.1 XM_011522181.2 XM_011522182.1 XM_011522183.2 NM_207581.3 XM_017022180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321437 c.855C>T p.S285S Chr.15: 45117801-45117801 on Build GRCh38
3672056 c.794G>T p.G265V Chr.15: 45117740-45117740 on Build GRCh38
6650208 c.1149C>T p.P383P Chr.15: 45117912-45117912 on Build GRCh38
6650206 c.1193G>A p.R398H Chr.15: 45117774-45117774 on Build GRCh38
3501657 c.899T>G p.L300R Chr.15: 45117845-45117845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761057646 Chr.15: 45117885-45117885 on Build GRCh38
rs755775190 Chr.15: 45117850-45117850 on Build GRCh38
rs756307526 Chr.15: 45117760-45117760 on Build GRCh38
rs771345227 Chr.15: 45117808-45117808 on Build GRCh38
rs769034135 Chr.15: 45117894-45117894 on Build GRCh38
rs148093420 Chr.15: 45117839-45117840 on Build GRCh38
rs775890393 Chr.15: 45117819-45117819 on Build GRCh38
rs764235764 Chr.15: 45117763-45117763 on Build GRCh38
rs746342250 Chr.15: 45117871-45117871 on Build GRCh38
rs575532094 Chr.15: 45117876-45117876 on Build GRCh38
rs780963242 Chr.15: 45117844-45117844 on Build GRCh38
rs774881100 Chr.15: 45117815-45117815 on Build GRCh38
rs374399422 Chr.15: 45117861-45117861 on Build GRCh38
rs112166546 Chr.15: 45117770-45117770 on Build GRCh38
rs761336230 Chr.15: 45117707-45117707 on Build GRCh38
rs774591035 Chr.15: 45117720-45117720 on Build GRCh38
rs1047116104 Chr.15: 45117805-45117805 on Build GRCh38
rs773750998 Chr.15: 45117709-45117709 on Build GRCh38
rs748779878 Chr.15: 45117852-45117852 on Build GRCh38
rs772628548 Chr.15: 45117873-45117873 on Build GRCh38
rs370420206 Chr.15: 45117851-45117851 on Build GRCh38
rs78064372 Chr.15: 45117706-45117706 on Build GRCh38
rs764323922 Chr.15: 45117824-45117824 on Build GRCh38
rs908649402 Chr.15: 45117857-45117857 on Build GRCh38
rs775982059 Chr.15: 45117880-45117880 on Build GRCh38
rs769363509 Chr.15: 45117791-45117791 on Build GRCh38
rs759877608 Chr.15: 45117726-45117726 on Build GRCh38
rs759731631 Chr.15: 45117805-45117805 on Build GRCh38
rs755589785 Chr.15: 45117818-45117818 on Build GRCh38
rs754808103 Chr.15: 45117842-45117842 on Build GRCh38
rs767881956 Chr.15: 45117727-45117727 on Build GRCh38
rs760971785 Chr.15: 45117823-45117823 on Build GRCh38
rs1057233703 Chr.15: 45117892-45117892 on Build GRCh38
rs747767492 Chr.15: 45117784-45117784 on Build GRCh38
rs752133621 Chr.15: 45117800-45117800 on Build GRCh38
rs910431511 Chr.15: 45117743-45117743 on Build GRCh38
rs751492604 Chr.15: 45117838-45117838 on Build GRCh38
rs750438741 Chr.15: 45117767-45117767 on Build GRCh38
rs751628515 Chr.15: 45117722-45117723 on Build GRCh38
rs769271781 Chr.15: 45117708-45117708 on Build GRCh38
rs766823135 Chr.15: 45117717-45117717 on Build GRCh38
rs772697087 Chr.15: 45117793-45117793 on Build GRCh38
rs753869388 Chr.15: 45117827-45117827 on Build GRCh38
rs759839746 Chr.15: 45117817-45117817 on Build GRCh38
rs201718435 Chr.15: 45117848-45117848 on Build GRCh38
rs770426058 Chr.15: 45117856-45117856 on Build GRCh38
rs746815709 Chr.15: 45117775-45117775 on Build GRCh38
rs752914323 Chr.15: 45117752-45117752 on Build GRCh38
rs754722228 Chr.15: 45117781-45117781 on Build GRCh38
rs763366208 Chr.15: 45117712-45117712 on Build GRCh38

View Full Product Details