Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240560980-240561229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 NM_001033575.1

Gene Symbol
DUSP28
Entrez Gene ID
285193
Gene Name
dual specificity phosphatase 28
Gene Aliases
DUSP26, VHP
UniGene
Hs.369297
Gene Chromosome Location
Chr.2: 240560054-240564014 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 NM_001033575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408627 c.319C>G p.R107G Chr.2: 240561003-240561003 on Build GRCh38
6408781 c.347C>A p.A116E Chr.2: 240561031-240561031 on Build GRCh38
5354243 c.379G>A p.A127T Chr.2: 240561063-240561063 on Build GRCh38
6157407 c.329C>T p.S110L Chr.2: 240561013-240561013 on Build GRCh38
3798812 c.312G>C p.K104N Chr.2: 240560996-240560996 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201793117 Chr.2: 240561039-240561039 on Build GRCh38
rs906236717 Chr.2: 240561152-240561152 on Build GRCh38
rs777055749 Chr.2: 240561063-240561063 on Build GRCh38
rs867347548 Chr.2: 240561028-240561028 on Build GRCh38
rs771443330 Chr.2: 240561101-240561101 on Build GRCh38
rs1011925554 Chr.2: 240561140-240561140 on Build GRCh38
rs571854171 Chr.2: 240561013-240561013 on Build GRCh38
rs542291327 Chr.2: 240561129-240561129 on Build GRCh38
rs982096102 Chr.2: 240561016-240561016 on Build GRCh38
rs1023226275 Chr.2: 240561051-240561051 on Build GRCh38
rs760226423 Chr.2: 240561121-240561121 on Build GRCh38
rs1016404757 Chr.2: 240561180-240561180 on Build GRCh38
rs886210827 Chr.2: 240561095-240561111 on Build GRCh38
rs575983369 Chr.2: 240561196-240561196 on Build GRCh38
rs758640402 Chr.2: 240561009-240561009 on Build GRCh38
rs74753884 Chr.2: 240561190-240561190 on Build GRCh38
rs1003154166 Chr.2: 240561153-240561153 on Build GRCh38
rs907902179 Chr.2: 240561019-240561019 on Build GRCh38
rs1036393246 Chr.2: 240561156-240561156 on Build GRCh38
rs765590102 Chr.2: 240561005-240561005 on Build GRCh38
rs571403854 Chr.2: 240561064-240561064 on Build GRCh38
rs973443326 Chr.2: 240561148-240561148 on Build GRCh38
rs769649960 Chr.2: 240561041-240561041 on Build GRCh38
rs754428063 Chr.2: 240561093-240561093 on Build GRCh38
rs780192561 Chr.2: 240561084-240561084 on Build GRCh38
rs772729836 Chr.2: 240561102-240561102 on Build GRCh38
rs537091038 Chr.2: 240561043-240561043 on Build GRCh38
rs983634963 Chr.2: 240561079-240561079 on Build GRCh38
rs933196842 Chr.2: 240561020-240561020 on Build GRCh38
rs780978886 Chr.2: 240561032-240561032 on Build GRCh38
rs752876567 Chr.2: 240561008-240561008 on Build GRCh38
rs1032543680 Chr.2: 240561040-240561040 on Build GRCh38
rs768174886 Chr.2: 240561049-240561049 on Build GRCh38
rs796295010 Chr.2: 240561075-240561091 on Build GRCh38
rs754471524 Chr.2: 240561133-240561133 on Build GRCh38
rs556419229 Chr.2: 240561048-240561048 on Build GRCh38
rs371287024 Chr.2: 240561134-240561134 on Build GRCh38
rs111241666 Chr.2: 240561147-240561147 on Build GRCh38
rs751867093 Chr.2: 240561076-240561076 on Build GRCh38
rs902421764 Chr.2: 240561099-240561099 on Build GRCh38
rs935202834 Chr.2: 240561127-240561127 on Build GRCh38
rs774117553 Chr.2: 240561053-240561053 on Build GRCh38
rs756146137 Chr.2: 240561083-240561083 on Build GRCh38
rs201249226 Chr.2: 240561074-240561074 on Build GRCh38
rs1039013938 Chr.2: 240561012-240561012 on Build GRCh38
rs781430049 Chr.2: 240561078-240561078 on Build GRCh38
rs920653506 Chr.2: 240561150-240561150 on Build GRCh38
rs889680518 Chr.2: 240561058-240561058 on Build GRCh38
rs530184793 Chr.2: 240561011-240561011 on Build GRCh38
rs538932275 Chr.2: 240561070-240561070 on Build GRCh38
rs755273079 Chr.2: 240561003-240561003 on Build GRCh38
rs1051616309 Chr.2: 240561030-240561030 on Build GRCh38
rs767063982 Chr.2: 240561062-240561062 on Build GRCh38
rs963475709 Chr.2: 240561205-240561205 on Build GRCh38
rs933290295 Chr.2: 240561093-240561093 on Build GRCh38
rs753036166 Chr.2: 240561073-240561073 on Build GRCh38
rs950957973 Chr.2: 240561170-240561170 on Build GRCh38
rs757425889 Chr.2: 240561025-240561025 on Build GRCh38
rs10933615 Chr.2: 240561091-240561091 on Build GRCh38
rs768550670 Chr.2: 240561085-240561085 on Build GRCh38
rs984033615 Chr.2: 240561172-240561172 on Build GRCh38
rs745764545 Chr.2: 240561082-240561082 on Build GRCh38
rs1023839799 Chr.2: 240561110-240561110 on Build GRCh38
rs766793961 Chr.2: 240561063-240561064 on Build GRCh38
rs761212547 Chr.2: 240561057-240561057 on Build GRCh38
rs768988257 Chr.2: 240561190-240561193 on Build GRCh38
rs747836528 Chr.2: 240561096-240561096 on Build GRCh38
rs767086994 Chr.2: 240561189-240561190 on Build GRCh38
rs1053588622 Chr.2: 240561128-240561128 on Build GRCh38
rs370148084 Chr.2: 240561069-240561069 on Build GRCh38
rs566287242 Chr.2: 240561010-240561010 on Build GRCh38
rs758876457 Chr.2: 240561025-240561043 on Build GRCh38
rs866357062 Chr.2: 240561038-240561038 on Build GRCh38
rs1015181376 Chr.2: 240561142-240561142 on Build GRCh38
rs751142968 Chr.2: 240561076-240561091 on Build GRCh38
rs750977042 Chr.2: 240561008-240561009 on Build GRCh38
rs974754904 Chr.2: 240561087-240561087 on Build GRCh38

View Full Product Details