Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28901135-28901404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS13
Entrez Gene ID
147015
Gene Name
dehydrogenase/reductase 13
Gene Aliases
SDR7C5
UniGene
Hs.631760
Gene Chromosome Location
Chr.17: 28897781-28903071 on Build GRCh38
Transcripts
NM_144683.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5385928 c.521G>C p.C174S Chr.17: 28901151-28901151 on Build GRCh38
977046 c.491G>A p.R164H Chr.17: 28901181-28901181 on Build GRCh38
5055334 c.395G>A p.R132H Chr.17: 28901277-28901277 on Build GRCh38
1302571 c.412C>T p.L138L Chr.17: 28901260-28901260 on Build GRCh38
5823333 c.536A>G p.D179G Chr.17: 28901136-28901136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911268551 Chr.17: 28901192-28901192 on Build GRCh38
rs769890272 Chr.17: 28901233-28901233 on Build GRCh38
rs755324575 Chr.17: 28901182-28901182 on Build GRCh38
rs147458712 Chr.17: 28901179-28901179 on Build GRCh38
rs149335962 Chr.17: 28901181-28901181 on Build GRCh38
rs962257038 Chr.17: 28901157-28901157 on Build GRCh38
rs767496229 Chr.17: 28901161-28901161 on Build GRCh38
rs953297242 Chr.17: 28901371-28901371 on Build GRCh38
rs867568916 Chr.17: 28901197-28901197 on Build GRCh38
rs751212616 Chr.17: 28901291-28901291 on Build GRCh38
rs766827886 Chr.17: 28901277-28901277 on Build GRCh38
rs199664494 Chr.17: 28901240-28901240 on Build GRCh38
rs913431290 Chr.17: 28901253-28901253 on Build GRCh38
rs749182871 Chr.17: 28901326-28901326 on Build GRCh38
rs771836972 Chr.17: 28901173-28901173 on Build GRCh38
rs755304833 Chr.17: 28901283-28901283 on Build GRCh38
rs538333570 Chr.17: 28901325-28901325 on Build GRCh38
rs762759248 Chr.17: 28901239-28901239 on Build GRCh38
rs756925355 Chr.17: 28901292-28901292 on Build GRCh38
rs145991183 Chr.17: 28901284-28901284 on Build GRCh38
rs761952621 Chr.17: 28901243-28901243 on Build GRCh38
rs533232643 Chr.17: 28901228-28901228 on Build GRCh38
rs761909064 Chr.17: 28901158-28901158 on Build GRCh38
rs34200761 Chr.17: 28901276-28901276 on Build GRCh38
rs568531176 Chr.17: 28901211-28901211 on Build GRCh38
rs777442522 Chr.17: 28901195-28901195 on Build GRCh38
rs780908843 Chr.17: 28901209-28901209 on Build GRCh38
rs774069075 Chr.17: 28901338-28901338 on Build GRCh38
rs755707381 Chr.17: 28901311-28901311 on Build GRCh38
rs772332173 Chr.17: 28901353-28901353 on Build GRCh38
rs750545604 Chr.17: 28901273-28901273 on Build GRCh38
rs746646326 Chr.17: 28901196-28901196 on Build GRCh38
rs368229450 Chr.17: 28901336-28901336 on Build GRCh38
rs201301706 Chr.17: 28901271-28901271 on Build GRCh38
rs4795474 Chr.17: 28901180-28901180 on Build GRCh38
rs148463808 Chr.17: 28901236-28901236 on Build GRCh38
rs770417842 Chr.17: 28901204-28901204 on Build GRCh38
rs373076605 Chr.17: 28901310-28901310 on Build GRCh38
rs953461222 Chr.17: 28901379-28901379 on Build GRCh38
rs751748998 Chr.17: 28901155-28901155 on Build GRCh38
rs748372545 Chr.17: 28901351-28901351 on Build GRCh38
rs551981860 Chr.17: 28901235-28901235 on Build GRCh38
rs745556247 Chr.17: 28901167-28901168 on Build GRCh38
rs774563934 Chr.17: 28901242-28901242 on Build GRCh38
rs1006823940 Chr.17: 28901374-28901374 on Build GRCh38
rs549688711 Chr.17: 28901270-28901270 on Build GRCh38
rs1030819110 Chr.17: 28901337-28901337 on Build GRCh38
rs1005998918 Chr.17: 28901304-28901304 on Build GRCh38
rs139866499 Chr.17: 28901300-28901300 on Build GRCh38
rs370018835 Chr.17: 28901306-28901306 on Build GRCh38
rs376526542 Chr.17: 28901278-28901278 on Build GRCh38

View Full Product Details