Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232567133-232567362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E2
Entrez Gene ID
9470
Gene Name
eukaryotic translation initiation factor 4E family member 2
Gene Aliases
4E-LP, 4EHP, EIF4EL3, IF4e
UniGene
Hs.292026
Gene Chromosome Location
Chr.2: 232550587-232583645 on Build GRCh38
Transcripts
NM_001276336.1 NM_001276337.1 NM_001282958.1 NM_004846.3 XM_005246975.3 XM_011512206.2 XM_017005377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3798752 c.606A>G p.R202R Chr.2: 232567155-232567155 on Build GRCh38
6652930 c.654C>A p.T218T Chr.2: 232567203-232567203 on Build GRCh38
1669851 c.604C>T p.R202* Chr.2: 232567153-232567153 on Build GRCh38
7131813 c.590G>T p.R197L Chr.2: 232567139-232567139 on Build GRCh38
1669852 c.645A>C p.K215N Chr.2: 232567194-232567194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748689451 Chr.2: 232567165-232567165 on Build GRCh38
rs184556365 Chr.2: 232567281-232567281 on Build GRCh38
rs778125062 Chr.2: 232567191-232567191 on Build GRCh38
rs765384219 Chr.2: 232567268-232567268 on Build GRCh38
rs377082781 Chr.2: 232567229-232567229 on Build GRCh38
rs937559279 Chr.2: 232567248-232567248 on Build GRCh38
rs1018223516 Chr.2: 232567300-232567300 on Build GRCh38
rs1030103516 Chr.2: 232567337-232567337 on Build GRCh38
rs1055983294 Chr.2: 232567259-232567259 on Build GRCh38
rs376551025 Chr.2: 232567261-232567261 on Build GRCh38
rs779332394 Chr.2: 232567159-232567159 on Build GRCh38
rs372575899 Chr.2: 232567225-232567225 on Build GRCh38
rs758911997 Chr.2: 232567275-232567275 on Build GRCh38
rs757618356 Chr.2: 232567287-232567287 on Build GRCh38
rs138511744 Chr.2: 232567297-232567297 on Build GRCh38
rs370502816 Chr.2: 232567232-232567232 on Build GRCh38
rs942224134 Chr.2: 232567270-232567270 on Build GRCh38
rs918117025 Chr.2: 232567204-232567204 on Build GRCh38
rs189317158 Chr.2: 232567290-232567290 on Build GRCh38
rs150475278 Chr.2: 232567203-232567203 on Build GRCh38
rs73101838 Chr.2: 232567278-232567278 on Build GRCh38
rs981504340 Chr.2: 232567231-232567231 on Build GRCh38
rs774289472 Chr.2: 232567249-232567249 on Build GRCh38
rs766560984 Chr.2: 232567256-232567256 on Build GRCh38
rs770558286 Chr.2: 232567219-232567219 on Build GRCh38
rs946030605 Chr.2: 232567221-232567221 on Build GRCh38
rs755118246 Chr.2: 232567262-232567262 on Build GRCh38
rs140247506 Chr.2: 232567327-232567327 on Build GRCh38
rs757329456 Chr.2: 232567218-232567218 on Build GRCh38
rs1037414454 Chr.2: 232567279-232567279 on Build GRCh38
rs764061891 Chr.2: 232567242-232567242 on Build GRCh38
rs752715459 Chr.2: 232567269-232567269 on Build GRCh38
rs1038513293 Chr.2: 232567274-232567274 on Build GRCh38
rs745565953 Chr.2: 232567292-232567292 on Build GRCh38
rs370036253 Chr.2: 232567179-232567179 on Build GRCh38
rs754063613 Chr.2: 232567257-232567257 on Build GRCh38
rs374564717 Chr.2: 232567241-232567241 on Build GRCh38
rs761693577 Chr.2: 232567250-232567250 on Build GRCh38
rs769278647 Chr.2: 232567296-232567296 on Build GRCh38

View Full Product Details