Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77425511-77425778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA14
Entrez Gene ID
9630
Gene Name
G protein subunit alpha 14
Gene Aliases
-
UniGene
Hs.657795
Gene Chromosome Location
Chr.9: 77423079-77648316 on Build GRCh38
Transcripts
NM_004297.3 XM_011519224.1 XM_017015341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1490140 c.809A>T p.N270I Chr.9: 77425630-77425630 on Build GRCh38
6672124 c.745G>A p.A249T Chr.9: 77425694-77425694 on Build GRCh38
1110315 c.727C>T p.R243C Chr.9: 77425712-77425712 on Build GRCh38
6013514 c.733G>A p.E245K Chr.9: 77425706-77425706 on Build GRCh38
5659328 c.731T>A p.M244K Chr.9: 77425708-77425708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747051087 Chr.9: 77425716-77425716 on Build GRCh38
rs757787418 Chr.9: 77425736-77425736 on Build GRCh38
rs749886105 Chr.9: 77425594-77425594 on Build GRCh38
rs369919018 Chr.9: 77425654-77425654 on Build GRCh38
rs1801259 Chr.9: 77425650-77425650 on Build GRCh38
rs751848300 Chr.9: 77425567-77425567 on Build GRCh38
rs746273540 Chr.9: 77425548-77425548 on Build GRCh38
rs572996758 Chr.9: 77425658-77425658 on Build GRCh38
rs759233048 Chr.9: 77425607-77425607 on Build GRCh38
rs753599450 Chr.9: 77425708-77425708 on Build GRCh38
rs200919606 Chr.9: 77425751-77425751 on Build GRCh38
rs897435968 Chr.9: 77425624-77425624 on Build GRCh38
rs532679687 Chr.9: 77425711-77425711 on Build GRCh38
rs563609547 Chr.9: 77425675-77425675 on Build GRCh38
rs771080036 Chr.9: 77425731-77425731 on Build GRCh38
rs778198432 Chr.9: 77425633-77425635 on Build GRCh38
rs868237297 Chr.9: 77425613-77425613 on Build GRCh38
rs750379315 Chr.9: 77425712-77425712 on Build GRCh38
rs79295433 Chr.9: 77425547-77425547 on Build GRCh38
rs201613861 Chr.9: 77425545-77425545 on Build GRCh38
rs556831126 Chr.9: 77425618-77425618 on Build GRCh38
rs758706347 Chr.9: 77425592-77425593 on Build GRCh38
rs780770982 Chr.9: 77425530-77425530 on Build GRCh38
rs1026965890 Chr.9: 77425629-77425629 on Build GRCh38
rs766501518 Chr.9: 77425561-77425561 on Build GRCh38
rs777218501 Chr.9: 77425539-77425539 on Build GRCh38
rs773830556 Chr.9: 77425651-77425651 on Build GRCh38
rs763838371 Chr.9: 77425679-77425679 on Build GRCh38
rs994560247 Chr.9: 77425626-77425626 on Build GRCh38
rs769089431 Chr.9: 77425633-77425633 on Build GRCh38
rs772468318 Chr.9: 77425592-77425592 on Build GRCh38
rs748312580 Chr.9: 77425584-77425584 on Build GRCh38
rs1001586126 Chr.9: 77425664-77425664 on Build GRCh38
rs536957030 Chr.9: 77425600-77425600 on Build GRCh38
rs371904055 Chr.9: 77425578-77425578 on Build GRCh38
rs760398367 Chr.9: 77425572-77425572 on Build GRCh38
rs61755085 Chr.9: 77425579-77425579 on Build GRCh38
rs149517927 Chr.9: 77425589-77425589 on Build GRCh38
rs200526690 Chr.9: 77425622-77425622 on Build GRCh38
rs994858315 Chr.9: 77425570-77425570 on Build GRCh38
rs28395814 Chr.9: 77425665-77425665 on Build GRCh38
rs759778715 Chr.9: 77425669-77425669 on Build GRCh38
rs781712782 Chr.9: 77425647-77425647 on Build GRCh38
rs762941986 Chr.9: 77425657-77425657 on Build GRCh38
rs199732364 Chr.9: 77425730-77425730 on Build GRCh38
rs762429615 Chr.9: 77425541-77425541 on Build GRCh38
rs763145469 Chr.9: 77425551-77425551 on Build GRCh38
rs142760663 Chr.9: 77425709-77425709 on Build GRCh38

View Full Product Details