Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3015620-3015893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
UniGene
Hs.274873
Gene Chromosome Location
Chr.11: 3000922-3057451 on Build GRCh38
Transcripts
NM_001014437.2 NM_001194997.1 NM_001751.5 NM_139273.3 NR_036542.1 XM_006718341.3 XM_017018389.1 XM_017018390.1 XM_017018391.1 XM_017018392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979560 c.1969C>T p.P657S Chr.11: 3015798-3015798 on Build GRCh38
6620301 c.1680C>T p.A560A Chr.11: 3015838-3015838 on Build GRCh38
3979561 c.1720C>T p.P574S Chr.11: 3015798-3015798 on Build GRCh38
4574309 c.1720G>A p.V574I Chr.11: 3015837-3015837 on Build GRCh38
5970141 c.1706T>A p.F569Y Chr.11: 3015812-3015812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004181992 Chr.11: 3015732-3015732 on Build GRCh38
rs753683711 Chr.11: 3015750-3015750 on Build GRCh38
rs553733646 Chr.11: 3015747-3015747 on Build GRCh38
rs779776886 Chr.11: 3015749-3015749 on Build GRCh38
rs979122976 Chr.11: 3015860-3015860 on Build GRCh38
rs114985608 Chr.11: 3015797-3015797 on Build GRCh38
rs376797594 Chr.11: 3015810-3015810 on Build GRCh38
rs372168614 Chr.11: 3015736-3015736 on Build GRCh38
rs963397018 Chr.11: 3015759-3015759 on Build GRCh38
rs988305307 Chr.11: 3015667-3015667 on Build GRCh38
rs377142673 Chr.11: 3015783-3015783 on Build GRCh38
rs543768913 Chr.11: 3015660-3015660 on Build GRCh38
rs946468675 Chr.11: 3015683-3015683 on Build GRCh38
rs960377474 Chr.11: 3015666-3015666 on Build GRCh38
rs898618586 Chr.11: 3015735-3015735 on Build GRCh38
rs763534078 Chr.11: 3015734-3015734 on Build GRCh38
rs770988521 Chr.11: 3015655-3015655 on Build GRCh38
rs373891738 Chr.11: 3015809-3015809 on Build GRCh38
rs1051263879 Chr.11: 3015727-3015727 on Build GRCh38
rs1010187259 Chr.11: 3015708-3015708 on Build GRCh38
rs932918704 Chr.11: 3015726-3015726 on Build GRCh38
rs774071174 Chr.11: 3015656-3015656 on Build GRCh38
rs750931201 Chr.11: 3015762-3015762 on Build GRCh38
rs535204333 Chr.11: 3015738-3015738 on Build GRCh38
rs767637585 Chr.11: 3015870-3015870 on Build GRCh38
rs571042655 Chr.11: 3015713-3015713 on Build GRCh38
rs770340203 Chr.11: 3015837-3015837 on Build GRCh38
rs1052828179 Chr.11: 3015682-3015682 on Build GRCh38
rs973388618 Chr.11: 3015786-3015786 on Build GRCh38
rs139930930 Chr.11: 3015790-3015790 on Build GRCh38
rs890584897 Chr.11: 3015718-3015718 on Build GRCh38
rs774594526 Chr.11: 3015861-3015861 on Build GRCh38
rs761855514 Chr.11: 3015867-3015867 on Build GRCh38
rs891122726 Chr.11: 3015728-3015728 on Build GRCh38
rs537154283 Chr.11: 3015668-3015668 on Build GRCh38
rs746431414 Chr.11: 3015832-3015832 on Build GRCh38
rs768720693 Chr.11: 3015848-3015848 on Build GRCh38
rs758967181 Chr.11: 3015804-3015804 on Build GRCh38
rs2071104 Chr.11: 3015703-3015703 on Build GRCh38
rs781761299 Chr.11: 3015812-3015812 on Build GRCh38
rs913686551 Chr.11: 3015673-3015673 on Build GRCh38
rs751913011 Chr.11: 3015828-3015828 on Build GRCh38
rs149736730 Chr.11: 3015791-3015791 on Build GRCh38
rs557908149 Chr.11: 3015702-3015702 on Build GRCh38
rs148488935 Chr.11: 3015838-3015838 on Build GRCh38
rs980668428 Chr.11: 3015648-3015648 on Build GRCh38
rs747352289 Chr.11: 3015859-3015859 on Build GRCh38

View Full Product Details