Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97037191-97037452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSV
Entrez Gene ID
1515
Gene Name
cathepsin V
Gene Aliases
CATL2, CTSL2, CTSU
UniGene
Hs.610096
Gene Chromosome Location
Chr.9: 97029677-97039643 on Build GRCh38
Transcripts
NM_001201575.1 NM_001333.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395945 c.396+2T>C p.? Chr.9: 97037250-97037250 on Build GRCh38
6900769 c.359G>T p.W120L Chr.9: 97037289-97037289 on Build GRCh38
7201791 c.282C>T p.C94C Chr.9: 97037366-97037366 on Build GRCh38
3996703 c.261A>C p.E87D Chr.9: 97037387-97037387 on Build GRCh38
3659902 c.286C>T p.R96* Chr.9: 97037362-97037362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768347914 Chr.9: 97037329-97037329 on Build GRCh38
rs374341587 Chr.9: 97037264-97037264 on Build GRCh38
rs267602328 Chr.9: 97037277-97037277 on Build GRCh38
rs754510545 Chr.9: 97037272-97037272 on Build GRCh38
rs532349961 Chr.9: 97037328-97037328 on Build GRCh38
rs187617973 Chr.9: 97037384-97037384 on Build GRCh38
rs768720614 Chr.9: 97037219-97037219 on Build GRCh38
rs369075309 Chr.9: 97037403-97037403 on Build GRCh38
rs770241197 Chr.9: 97037361-97037361 on Build GRCh38
rs747111038 Chr.9: 97037218-97037218 on Build GRCh38
rs896342676 Chr.9: 97037357-97037357 on Build GRCh38
rs199884373 Chr.9: 97037241-97037241 on Build GRCh38
rs368226558 Chr.9: 97037380-97037380 on Build GRCh38
rs745962692 Chr.9: 97037343-97037343 on Build GRCh38
rs139348613 Chr.9: 97037279-97037279 on Build GRCh38
rs749091158 Chr.9: 97037282-97037282 on Build GRCh38
rs145383604 Chr.9: 97037376-97037376 on Build GRCh38
rs757134245 Chr.9: 97037291-97037291 on Build GRCh38
rs964069806 Chr.9: 97037336-97037336 on Build GRCh38
rs371288973 Chr.9: 97037421-97037421 on Build GRCh38
rs763754475 Chr.9: 97037391-97037391 on Build GRCh38
rs778852809 Chr.9: 97037310-97037310 on Build GRCh38
rs150068143 Chr.9: 97037319-97037319 on Build GRCh38
rs372492323 Chr.9: 97037244-97037244 on Build GRCh38
rs774981893 Chr.9: 97037361-97037361 on Build GRCh38
rs753586971 Chr.9: 97037394-97037394 on Build GRCh38
rs773663582 Chr.9: 97037339-97037339 on Build GRCh38
rs763498015 Chr.9: 97037267-97037267 on Build GRCh38
rs559045548 Chr.9: 97037311-97037311 on Build GRCh38
rs775486161 Chr.9: 97037217-97037217 on Build GRCh38
rs760021696 Chr.9: 97037371-97037371 on Build GRCh38
rs375981795 Chr.9: 97037399-97037399 on Build GRCh38
rs763442831 Chr.9: 97037347-97037347 on Build GRCh38
rs1019686442 Chr.9: 97037335-97037335 on Build GRCh38
rs761989224 Chr.9: 97037242-97037242 on Build GRCh38
rs752383893 Chr.9: 97037275-97037275 on Build GRCh38
rs759378406 Chr.9: 97037405-97037405 on Build GRCh38
rs1023394620 Chr.9: 97037362-97037362 on Build GRCh38
rs751381692 Chr.9: 97037273-97037273 on Build GRCh38
rs776630762 Chr.9: 97037322-97037322 on Build GRCh38
rs766894046 Chr.9: 97037351-97037351 on Build GRCh38
rs751076789 Chr.9: 97037268-97037268 on Build GRCh38

View Full Product Details