Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28993314-28993583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7136538 c.608C>A p.P203Q Chr.1: 28993469-28993469 on Build GRCh38
3487830 c.518G>A p.G173E Chr.1: 28993379-28993379 on Build GRCh38
1341619 c.656_657delAC p.T220fs*3 Chr.1: 28993517-28993518 on Build GRCh38
6581090 c.601C>G p.Q201E Chr.1: 28993462-28993462 on Build GRCh38
7273948 c.625A>G p.N209D Chr.1: 28993486-28993486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748799779 Chr.1: 28993467-28993467 on Build GRCh38
rs778639972 Chr.1: 28993543-28993546 on Build GRCh38
rs147738349 Chr.1: 28993349-28993349 on Build GRCh38
rs957128141 Chr.1: 28993482-28993482 on Build GRCh38
rs775969811 Chr.1: 28993348-28993348 on Build GRCh38
rs769005503 Chr.1: 28993513-28993513 on Build GRCh38
rs754849107 Chr.1: 28993399-28993399 on Build GRCh38
rs777346153 Chr.1: 28993445-28993445 on Build GRCh38
rs111642750 Chr.1: 28993501-28993501 on Build GRCh38
rs767424892 Chr.1: 28993549-28993549 on Build GRCh38
rs759585466 Chr.1: 28993545-28993545 on Build GRCh38
rs753982388 Chr.1: 28993382-28993382 on Build GRCh38
rs756894056 Chr.1: 28993415-28993417 on Build GRCh38
rs866071277 Chr.1: 28993405-28993405 on Build GRCh38
rs751463925 Chr.1: 28993397-28993397 on Build GRCh38
rs747929575 Chr.1: 28993415-28993415 on Build GRCh38
rs750437420 Chr.1: 28993540-28993540 on Build GRCh38
rs755818196 Chr.1: 28993439-28993439 on Build GRCh38
rs557047271 Chr.1: 28993404-28993404 on Build GRCh38
rs146497501 Chr.1: 28993539-28993539 on Build GRCh38
rs761064131 Chr.1: 28993507-28993507 on Build GRCh38
rs866390552 Chr.1: 28993456-28993456 on Build GRCh38
rs778268635 Chr.1: 28993491-28993491 on Build GRCh38
rs573494559 Chr.1: 28993366-28993366 on Build GRCh38
rs148968117 Chr.1: 28993522-28993522 on Build GRCh38
rs781004502 Chr.1: 28993401-28993401 on Build GRCh38
rs753385565 Chr.1: 28993377-28993379 on Build GRCh38
rs573738310 Chr.1: 28993479-28993479 on Build GRCh38
rs766257622 Chr.1: 28993393-28993393 on Build GRCh38
rs757297069 Chr.1: 28993390-28993390 on Build GRCh38
rs776954685 Chr.1: 28993519-28993519 on Build GRCh38
rs368577557 Chr.1: 28993497-28993497 on Build GRCh38
rs1032686907 Chr.1: 28993463-28993463 on Build GRCh38
rs746430671 Chr.1: 28993496-28993496 on Build GRCh38
rs201231112 Chr.1: 28993381-28993381 on Build GRCh38
rs121434564 Chr.1: 28993490-28993490 on Build GRCh38

View Full Product Details