Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197090689-197090948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPM
Entrez Gene ID
259266
Gene Name
abnormal spindle microtubule assembly
Gene Aliases
ASP, Calmbp1, MCPH5
UniGene
Hs.121028
Gene Chromosome Location
Chr.1: 197084127-197146694 on Build GRCh38
Transcripts
NM_001206846.1 NM_018136.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901645 c.9577C>T p.R3193C Chr.1: 197090909-197090909 on Build GRCh38
358309 c.9616A>G p.S3206G Chr.1: 197090870-197090870 on Build GRCh38
238861 c.9546T>A p.N3182K Chr.1: 197090940-197090940 on Build GRCh38
3481010 c.9592C>T p.R3198C Chr.1: 197090894-197090894 on Build GRCh38
1718102 c.9584T>A p.F3195Y Chr.1: 197090902-197090902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957152605 Chr.1: 197090898-197090898 on Build GRCh38
rs778653050 Chr.1: 197090869-197090869 on Build GRCh38
rs967942722 Chr.1: 197090882-197090882 on Build GRCh38
rs4915316 Chr.1: 197090798-197090798 on Build GRCh38
rs766410220 Chr.1: 197090805-197090805 on Build GRCh38
rs772021575 Chr.1: 197090738-197090738 on Build GRCh38
rs768731480 Chr.1: 197090862-197090862 on Build GRCh38
rs201186097 Chr.1: 197090782-197090782 on Build GRCh38
rs140679756 Chr.1: 197090909-197090909 on Build GRCh38
rs937993437 Chr.1: 197090769-197090769 on Build GRCh38
rs376130761 Chr.1: 197090850-197090850 on Build GRCh38
rs1001011460 Chr.1: 197090834-197090834 on Build GRCh38
rs373418682 Chr.1: 197090908-197090908 on Build GRCh38
rs10754214 Chr.1: 197090762-197090762 on Build GRCh38
rs576658221 Chr.1: 197090732-197090732 on Build GRCh38
rs145333987 Chr.1: 197090893-197090893 on Build GRCh38
rs751806531 Chr.1: 197090801-197090801 on Build GRCh38
rs138624150 Chr.1: 197090922-197090922 on Build GRCh38
rs756010255 Chr.1: 197090807-197090807 on Build GRCh38
rs199422189 Chr.1: 197090929-197090929 on Build GRCh38
rs896689246 Chr.1: 197090784-197090784 on Build GRCh38
rs886044195 Chr.1: 197090913-197090913 on Build GRCh38
rs753092350 Chr.1: 197090917-197090917 on Build GRCh38
rs760292134 Chr.1: 197090894-197090894 on Build GRCh38
rs765801359 Chr.1: 197090915-197090915 on Build GRCh38
rs749298378 Chr.1: 197090827-197090827 on Build GRCh38
rs915309751 Chr.1: 197090828-197090828 on Build GRCh38
rs1038451612 Chr.1: 197090739-197090739 on Build GRCh38
rs199422190 Chr.1: 197090891-197090891 on Build GRCh38
rs751489737 Chr.1: 197090822-197090823 on Build GRCh38
rs763141390 Chr.1: 197090921-197090921 on Build GRCh38
rs4915315 Chr.1: 197090748-197090748 on Build GRCh38
rs1055134859 Chr.1: 197090783-197090783 on Build GRCh38
rs72736482 Chr.1: 197090734-197090734 on Build GRCh38
rs149351720 Chr.1: 197090742-197090742 on Build GRCh38
rs779988401 Chr.1: 197090808-197090808 on Build GRCh38
rs754852879 Chr.1: 197090836-197090837 on Build GRCh38

View Full Product Details