Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45649990-45650263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST1
Entrez Gene ID
8534
Gene Name
carbohydrate sulfotransferase 1
Gene Aliases
C6ST, GST-1, KS6ST, KSGal6ST, KSST
UniGene
Hs.104576
Gene Chromosome Location
Chr.11: 45647689-45665656 on Build GRCh38
Transcripts
NM_003654.5 XM_006718356.3 XM_017018459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5431032 c.728G>A p.R243H Chr.11: 45650196-45650196 on Build GRCh38
371691 c.813G>T p.Q271H Chr.11: 45650111-45650111 on Build GRCh38
403334 c.861C>T p.L287L Chr.11: 45650063-45650063 on Build GRCh38
6628822 c.780C>T p.T260T Chr.11: 45650144-45650144 on Build GRCh38
6025151 c.806T>C p.V269A Chr.11: 45650118-45650118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774865102 Chr.11: 45650161-45650161 on Build GRCh38
rs1028976256 Chr.11: 45650109-45650109 on Build GRCh38
rs769686602 Chr.11: 45650193-45650193 on Build GRCh38
rs768248608 Chr.11: 45650170-45650170 on Build GRCh38
rs111740567 Chr.11: 45650198-45650198 on Build GRCh38
rs772527405 Chr.11: 45650150-45650150 on Build GRCh38
rs749684408 Chr.11: 45650133-45650133 on Build GRCh38
rs139813023 Chr.11: 45650132-45650132 on Build GRCh38
rs377425314 Chr.11: 45650047-45650047 on Build GRCh38
rs976283351 Chr.11: 45650068-45650068 on Build GRCh38
rs965915401 Chr.11: 45650053-45650053 on Build GRCh38
rs745422470 Chr.11: 45650024-45650024 on Build GRCh38
rs752781504 Chr.11: 45650223-45650223 on Build GRCh38
rs761653590 Chr.11: 45650075-45650075 on Build GRCh38
rs746469231 Chr.11: 45650169-45650169 on Build GRCh38
rs771549271 Chr.11: 45650026-45650026 on Build GRCh38
rs757586414 Chr.11: 45650141-45650141 on Build GRCh38
rs1006152663 Chr.11: 45650192-45650192 on Build GRCh38
rs774931521 Chr.11: 45650032-45650032 on Build GRCh38
rs765327530 Chr.11: 45650076-45650076 on Build GRCh38
rs754698246 Chr.11: 45650095-45650095 on Build GRCh38
rs147428436 Chr.11: 45650103-45650103 on Build GRCh38
rs868485499 Chr.11: 45650059-45650059 on Build GRCh38
rs369014915 Chr.11: 45650102-45650102 on Build GRCh38
rs776340776 Chr.11: 45650174-45650174 on Build GRCh38
rs754069806 Chr.11: 45650234-45650234 on Build GRCh38
rs779445818 Chr.11: 45650144-45650144 on Build GRCh38
rs776300953 Chr.11: 45650056-45650056 on Build GRCh38
rs201148963 Chr.11: 45650144-45650154 on Build GRCh38
rs763733732 Chr.11: 45650052-45650052 on Build GRCh38
rs913862316 Chr.11: 45650073-45650073 on Build GRCh38
rs752757824 Chr.11: 45650115-45650115 on Build GRCh38
rs756306640 Chr.11: 45650119-45650119 on Build GRCh38
rs764265419 Chr.11: 45650232-45650232 on Build GRCh38
rs766364485 Chr.11: 45650206-45650206 on Build GRCh38
rs761549838 Chr.11: 45650177-45650177 on Build GRCh38
rs200408664 Chr.11: 45650225-45650225 on Build GRCh38

View Full Product Details