Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57125771-57126044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
UniGene
Hs.339809
Gene Chromosome Location
Chr.16: 57092543-57147966 on Build GRCh38
Transcripts
NM_152727.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510465 c.1021G>A p.A341T Chr.16: 57125953-57125953 on Build GRCh38
971800 c.981G>T p.L327F Chr.16: 57125913-57125913 on Build GRCh38
3510464 c.969C>T p.D323D Chr.16: 57125901-57125901 on Build GRCh38
2836291 c.1056C>T p.Y352Y Chr.16: 57125988-57125988 on Build GRCh38
1378537 c.954C>T p.N318N Chr.16: 57125886-57125886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750488322 Chr.16: 57125833-57125833 on Build GRCh38
rs745464905 Chr.16: 57126013-57126013 on Build GRCh38
rs767721335 Chr.16: 57125970-57125970 on Build GRCh38
rs770696614 Chr.16: 57125902-57125902 on Build GRCh38
rs990803257 Chr.16: 57125890-57125890 on Build GRCh38
rs150252579 Chr.16: 57125920-57125920 on Build GRCh38
rs751339795 Chr.16: 57125826-57125826 on Build GRCh38
rs553796398 Chr.16: 57125992-57125992 on Build GRCh38
rs201563361 Chr.16: 57125929-57125929 on Build GRCh38
rs756253825 Chr.16: 57125834-57125834 on Build GRCh38
rs774108311 Chr.16: 57125963-57125963 on Build GRCh38
rs751041768 Chr.16: 57125828-57125828 on Build GRCh38
rs748574324 Chr.16: 57125899-57125899 on Build GRCh38
rs184236869 Chr.16: 57125832-57125832 on Build GRCh38
rs750576421 Chr.16: 57125973-57125973 on Build GRCh38
rs774881469 Chr.16: 57125940-57125940 on Build GRCh38
rs775032124 Chr.16: 57126020-57126020 on Build GRCh38
rs762904697 Chr.16: 57125949-57125949 on Build GRCh38
rs368278934 Chr.16: 57125813-57125813 on Build GRCh38
rs758773496 Chr.16: 57126007-57126007 on Build GRCh38
rs766442892 Chr.16: 57125989-57125989 on Build GRCh38
rs755472444 Chr.16: 57125886-57125886 on Build GRCh38
rs563378631 Chr.16: 57125898-57125898 on Build GRCh38
rs763898029 Chr.16: 57125821-57125821 on Build GRCh38
rs761653124 Chr.16: 57125967-57125967 on Build GRCh38
rs377664960 Chr.16: 57126009-57126009 on Build GRCh38
rs758981413 Chr.16: 57125876-57125876 on Build GRCh38
rs151128903 Chr.16: 57125883-57125883 on Build GRCh38
rs780011907 Chr.16: 57125838-57125838 on Build GRCh38
rs779348766 Chr.16: 57125995-57125995 on Build GRCh38
rs781518184 Chr.16: 57125952-57125952 on Build GRCh38
rs755490883 Chr.16: 57125993-57125993 on Build GRCh38
rs763976532 Chr.16: 57125956-57125956 on Build GRCh38
rs375002363 Chr.16: 57126018-57126018 on Build GRCh38
rs763687706 Chr.16: 57125817-57125817 on Build GRCh38
rs375301498 Chr.16: 57125988-57125988 on Build GRCh38
rs748521195 Chr.16: 57125999-57125999 on Build GRCh38

View Full Product Details