Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45621567-45621810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
UniGene
Hs.18912
Gene Chromosome Location
Chr.1: 45620044-45624787 on Build GRCh38
Transcripts
NM_001114938.2 NM_001190182.1 XM_011540812.1 XM_011540814.1 XM_011540820.1 XM_017000450.1 XM_017000451.1 XM_017000452.1 XM_017000453.1 XM_017000454.1 XM_017000455.1 XM_017000456.1 XM_017000457.1 XM_017000458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562254 c.992C>G p.P331R Chr.1: 45621734-45621734 on Build GRCh38
5618178 c.1057G>A p.D353N Chr.1: 45621669-45621669 on Build GRCh38
910206 c.1036C>T p.H346Y Chr.1: 45621690-45621690 on Build GRCh38
4008529 c.1069C>T p.P357S Chr.1: 45621657-45621657 on Build GRCh38
4460857 c.1085C>T p.P362L Chr.1: 45621641-45621641 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375821787 Chr.1: 45621749-45621749 on Build GRCh38
rs775853873 Chr.1: 45621773-45621773 on Build GRCh38
rs747568235 Chr.1: 45621660-45621660 on Build GRCh38
rs906142707 Chr.1: 45621672-45621672 on Build GRCh38
rs775729215 Chr.1: 45621674-45621674 on Build GRCh38
rs373781070 Chr.1: 45621618-45621618 on Build GRCh38
rs779540649 Chr.1: 45621758-45621758 on Build GRCh38
rs779914863 Chr.1: 45621651-45621651 on Build GRCh38
rs750314777 Chr.1: 45621703-45621703 on Build GRCh38
rs753766451 Chr.1: 45621629-45621629 on Build GRCh38
rs760862274 Chr.1: 45621682-45621682 on Build GRCh38
rs779787845 Chr.1: 45621591-45621591 on Build GRCh38
rs772780495 Chr.1: 45621715-45621716 on Build GRCh38
rs752903141 Chr.1: 45621623-45621623 on Build GRCh38
rs772421315 Chr.1: 45621671-45621671 on Build GRCh38
rs778990176 Chr.1: 45621713-45621713 on Build GRCh38
rs773925529 Chr.1: 45621610-45621610 on Build GRCh38
rs901585011 Chr.1: 45621780-45621780 on Build GRCh38
rs776659421 Chr.1: 45621784-45621784 on Build GRCh38
rs375531461 Chr.1: 45621691-45621691 on Build GRCh38
rs201021595 Chr.1: 45621641-45621641 on Build GRCh38
rs766250058 Chr.1: 45621712-45621712 on Build GRCh38
rs764249753 Chr.1: 45621684-45621684 on Build GRCh38
rs764126888 Chr.1: 45621628-45621628 on Build GRCh38
rs769284177 Chr.1: 45621682-45621691 on Build GRCh38
rs780733912 Chr.1: 45621659-45621659 on Build GRCh38
rs755562243 Chr.1: 45621752-45621752 on Build GRCh38
rs772518775 Chr.1: 45621616-45621620 on Build GRCh38
rs774658907 Chr.1: 45621670-45621670 on Build GRCh38
rs760810886 Chr.1: 45621626-45621626 on Build GRCh38
rs772472177 Chr.1: 45621772-45621772 on Build GRCh38
rs1044565190 Chr.1: 45621649-45621649 on Build GRCh38
rs774807193 Chr.1: 45621614-45621614 on Build GRCh38
rs768272296 Chr.1: 45621599-45621599 on Build GRCh38
rs371538924 Chr.1: 45621722-45621722 on Build GRCh38
rs765234623 Chr.1: 45621698-45621698 on Build GRCh38
rs757243591 Chr.1: 45621631-45621631 on Build GRCh38
rs762457232 Chr.1: 45621769-45621770 on Build GRCh38
rs368751626 Chr.1: 45621696-45621696 on Build GRCh38
rs369312257 Chr.1: 45621776-45621776 on Build GRCh38
rs746532056 Chr.1: 45621594-45621594 on Build GRCh38
rs780394352 Chr.1: 45621629-45621629 on Build GRCh38
rs367850607 Chr.1: 45621648-45621648 on Build GRCh38
rs759749100 Chr.1: 45621616-45621616 on Build GRCh38
rs750238354 Chr.1: 45621647-45621647 on Build GRCh38
rs768872983 Chr.1: 45621782-45621782 on Build GRCh38
rs746172322 Chr.1: 45621609-45621610 on Build GRCh38
rs771470577 Chr.1: 45621669-45621669 on Build GRCh38
rs771290388 Chr.1: 45621613-45621613 on Build GRCh38
rs747601492 Chr.1: 45621649-45621649 on Build GRCh38
rs747703395 Chr.1: 45621751-45621751 on Build GRCh38
rs746343137 Chr.1: 45621767-45621767 on Build GRCh38
rs754706584 Chr.1: 45621738-45621738 on Build GRCh38
rs768325251 Chr.1: 45621656-45621656 on Build GRCh38
rs763488200 Chr.1: 45621612-45621612 on Build GRCh38

View Full Product Details