Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87179492-87179747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTF1
Entrez Gene ID
9988
Gene Name
cyclin D binding myb like transcription factor 1
Gene Aliases
DMP1, DMTF, MRUL, hDMP1
UniGene
Hs.196129
Gene Chromosome Location
Chr.7: 87152361-87196332 on Build GRCh38
Transcripts
NM_001142326.1 NM_001142327.1 NM_021145.3 NR_024549.1 NR_024550.1 XM_011516735.2 XM_011516737.1 XM_011516739.2 XM_017012865.1 XM_017012866.1 XM_017012867.1 XM_017012868.1 XM_017012869.1 XM_017012870.1 XM_017012871.1 XM_017012872.1 XM_017012873.1 XM_017012874.1 XM_017012875.1 XM_017012876.1 XM_017012877.1 XM_017012878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6568999 c.552C>G p.I184M Chr.7: 87179577-87179577 on Build GRCh38
6504651 c.614G>A p.R205Q Chr.7: 87179639-87179639 on Build GRCh38
6568998 c.552C>G p.I184M Chr.7: 87179577-87179577 on Build GRCh38
5641566 c.595A>G p.I199V Chr.7: 87179620-87179620 on Build GRCh38
6504650 c.614G>A p.R205Q Chr.7: 87179639-87179639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184925730 Chr.7: 87179595-87179595 on Build GRCh38
rs754637753 Chr.7: 87179711-87179711 on Build GRCh38
rs368478182 Chr.7: 87179655-87179655 on Build GRCh38
rs1001371900 Chr.7: 87179571-87179571 on Build GRCh38
rs754550499 Chr.7: 87179532-87179532 on Build GRCh38
rs537780294 Chr.7: 87179550-87179550 on Build GRCh38
rs751271276 Chr.7: 87179697-87179697 on Build GRCh38
rs773012634 Chr.7: 87179652-87179652 on Build GRCh38
rs746081360 Chr.7: 87179582-87179582 on Build GRCh38
rs774646752 Chr.7: 87179572-87179572 on Build GRCh38
rs368726048 Chr.7: 87179548-87179548 on Build GRCh38
rs574217533 Chr.7: 87179625-87179625 on Build GRCh38
rs778033879 Chr.7: 87179551-87179551 on Build GRCh38
rs375204119 Chr.7: 87179558-87179558 on Build GRCh38
rs751075195 Chr.7: 87179527-87179527 on Build GRCh38
rs897092873 Chr.7: 87179530-87179531 on Build GRCh38
rs771295078 Chr.7: 87179566-87179566 on Build GRCh38
rs762676722 Chr.7: 87179671-87179671 on Build GRCh38
rs766158853 Chr.7: 87179673-87179673 on Build GRCh38
rs766980732 Chr.7: 87179716-87179716 on Build GRCh38
rs140053946 Chr.7: 87179549-87179549 on Build GRCh38
rs761527606 Chr.7: 87179656-87179676 on Build GRCh38
rs761696960 Chr.7: 87179632-87179632 on Build GRCh38
rs765181504 Chr.7: 87179643-87179643 on Build GRCh38

View Full Product Details