Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183270099-183270372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT5
Entrez Gene ID
84002
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases
B3GN-T5, beta3Gn-T5
UniGene
Hs.718506
Gene Chromosome Location
Chr.3: 183253207-183273391 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

Gene Symbol
MCF2L2
Entrez Gene ID
23101
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Gene Aliases
ARHGEF22
UniGene
Hs.208267
Gene Chromosome Location
Chr.3: 183176116-183428990 on Build GRCh38
Transcripts
NM_032047.4 XM_005247823.4 XM_005247824.3 XM_005247825.3 XM_011513225.2 XM_011513226.2 XM_011513227.2 XM_011513228.2 XM_011513229.2 XM_017007309.1 NM_015078.3 XM_011512585.2 XM_011512586.2 XM_017005943.1 XM_017005945.1 XM_017005946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373098 c.533G>A p.W178* Chr.3: 183270331-183270331 on Build GRCh38
5026084 c.490T>A p.S164T Chr.3: 183270288-183270288 on Build GRCh38
337846 c.369C>T p.A123A Chr.3: 183270167-183270167 on Build GRCh38
3206587 c.553C>T p.H185Y Chr.3: 183270351-183270351 on Build GRCh38
4831553 c.451C>G p.Q151E Chr.3: 183270249-183270249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768307976 Chr.3: 183270336-183270336 on Build GRCh38
rs748325325 Chr.3: 183270137-183270137 on Build GRCh38
rs1049068337 Chr.3: 183270135-183270135 on Build GRCh38
rs142129979 Chr.3: 183270344-183270344 on Build GRCh38
rs755203514 Chr.3: 183270134-183270134 on Build GRCh38
rs747782074 Chr.3: 183270343-183270343 on Build GRCh38
rs767733409 Chr.3: 183270229-183270229 on Build GRCh38
rs180964101 Chr.3: 183270334-183270334 on Build GRCh38
rs565666641 Chr.3: 183270141-183270141 on Build GRCh38
rs781255035 Chr.3: 183270136-183270136 on Build GRCh38
rs752838845 Chr.3: 183270249-183270249 on Build GRCh38
rs769405398 Chr.3: 183270232-183270232 on Build GRCh38
rs747306979 Chr.3: 183270319-183270319 on Build GRCh38
rs1039538753 Chr.3: 183270160-183270160 on Build GRCh38
rs957615603 Chr.3: 183270292-183270292 on Build GRCh38
rs768413971 Chr.3: 183270181-183270181 on Build GRCh38
rs745821925 Chr.3: 183270150-183270150 on Build GRCh38
rs751976718 Chr.3: 183270228-183270228 on Build GRCh38
rs146160618 Chr.3: 183270165-183270165 on Build GRCh38
rs756310252 Chr.3: 183270265-183270265 on Build GRCh38
rs780423909 Chr.3: 183270128-183270128 on Build GRCh38
rs1002571616 Chr.3: 183270257-183270257 on Build GRCh38
rs749889616 Chr.3: 183270204-183270204 on Build GRCh38
rs369857642 Chr.3: 183270298-183270298 on Build GRCh38
rs759746825 Chr.3: 183270222-183270222 on Build GRCh38
rs781516085 Chr.3: 183270236-183270236 on Build GRCh38
rs764635314 Chr.3: 183270199-183270199 on Build GRCh38
rs568355269 Chr.3: 183270138-183270138 on Build GRCh38
rs201281355 Chr.3: 183270210-183270210 on Build GRCh38
rs778958256 Chr.3: 183270147-183270147 on Build GRCh38
rs140197372 Chr.3: 183270329-183270329 on Build GRCh38
rs111341243 Chr.3: 183270207-183270207 on Build GRCh38
rs776397039 Chr.3: 183270338-183270338 on Build GRCh38
rs755289892 Chr.3: 183270230-183270230 on Build GRCh38
rs751850381 Chr.3: 183270132-183270132 on Build GRCh38
rs761428945 Chr.3: 183270189-183270189 on Build GRCh38
rs902915427 Chr.3: 183270237-183270237 on Build GRCh38
rs754121621 Chr.3: 183270214-183270214 on Build GRCh38
rs760491067 Chr.3: 183270169-183270169 on Build GRCh38
rs776147136 Chr.3: 183270188-183270188 on Build GRCh38
rs373017855 Chr.3: 183270319-183270319 on Build GRCh38
rs772096430 Chr.3: 183270151-183270151 on Build GRCh38
rs758815965 Chr.3: 183270127-183270127 on Build GRCh38
rs1035756136 Chr.3: 183270281-183270281 on Build GRCh38
rs777744768 Chr.3: 183270283-183270283 on Build GRCh38

View Full Product Details