Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102930380-102930609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
UniGene
Hs.534494
Gene Chromosome Location
Chr.14: 102922430-102930842 on Build GRCh38
Transcripts
NM_030943.3 XM_011537202.2 XM_011537203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5950358 c.1267C>T p.R423W Chr.14: 102930585-102930585 on Build GRCh38
6434000 c.1171T>A p.W391R Chr.14: 102930407-102930407 on Build GRCh38
6421666 c.1217T>C p.F406S Chr.14: 102930453-102930453 on Build GRCh38
5777055 c.1221C>G p.R407R Chr.14: 102930457-102930457 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761239185 Chr.14: 102930558-102930558 on Build GRCh38
rs565796359 Chr.14: 102930406-102930406 on Build GRCh38
rs932137637 Chr.14: 102930403-102930403 on Build GRCh38
rs753700673 Chr.14: 102930586-102930586 on Build GRCh38
rs754968366 Chr.14: 102930511-102930511 on Build GRCh38
rs915321774 Chr.14: 102930528-102930528 on Build GRCh38
rs1003560283 Chr.14: 102930574-102930574 on Build GRCh38
rs1054299900 Chr.14: 102930519-102930519 on Build GRCh38
rs763394962 Chr.14: 102930546-102930546 on Build GRCh38
rs386834166 Chr.14: 102930503-102930503 on Build GRCh38
rs749049449 Chr.14: 102930509-102930509 on Build GRCh38
rs778907056 Chr.14: 102930513-102930513 on Build GRCh38
rs894558374 Chr.14: 102930435-102930435 on Build GRCh38
rs1030964062 Chr.14: 102930483-102930483 on Build GRCh38
rs912035219 Chr.14: 102930569-102930569 on Build GRCh38
rs1055765386 Chr.14: 102930432-102930432 on Build GRCh38
rs779783437 Chr.14: 102930491-102930491 on Build GRCh38
rs1012051558 Chr.14: 102930437-102930437 on Build GRCh38
rs750073599 Chr.14: 102930568-102930568 on Build GRCh38
rs776745054 Chr.14: 102930529-102930529 on Build GRCh38
rs899163860 Chr.14: 102930427-102930427 on Build GRCh38
rs754703477 Chr.14: 102930587-102930587 on Build GRCh38
rs534440441 Chr.14: 102930419-102930419 on Build GRCh38
rs746050993 Chr.14: 102930532-102930532 on Build GRCh38
rs757315069 Chr.14: 102930590-102930590 on Build GRCh38
rs755801781 Chr.14: 102930577-102930577 on Build GRCh38
rs1016598695 Chr.14: 102930433-102930433 on Build GRCh38
rs770113617 Chr.14: 102930533-102930533 on Build GRCh38
rs765992885 Chr.14: 102930581-102930581 on Build GRCh38
rs774850532 Chr.14: 102930557-102930557 on Build GRCh38
rs1052377523 Chr.14: 102930412-102930412 on Build GRCh38
rs962442161 Chr.14: 102930447-102930447 on Build GRCh38
rs748069971 Chr.14: 102930589-102930589 on Build GRCh38
rs34081720 Chr.14: 102930535-102930535 on Build GRCh38
rs759269975 Chr.14: 102930507-102930508 on Build GRCh38
rs374163665 Chr.14: 102930475-102930475 on Build GRCh38
rs924933674 Chr.14: 102930567-102930567 on Build GRCh38
rs912125076 Chr.14: 102930413-102930413 on Build GRCh38
rs994837555 Chr.14: 102930429-102930429 on Build GRCh38
rs940902893 Chr.14: 102930576-102930576 on Build GRCh38
rs1025245635 Chr.14: 102930470-102930470 on Build GRCh38
rs770986064 Chr.14: 102930517-102930517 on Build GRCh38
rs386834164 Chr.14: 102930400-102930400 on Build GRCh38
rs386834165 Chr.14: 102930489-102930490 on Build GRCh38
rs868529754 Chr.14: 102930456-102930456 on Build GRCh38
rs764761226 Chr.14: 102930547-102930547 on Build GRCh38
rs767004707 Chr.14: 102930562-102930562 on Build GRCh38
rs999834788 Chr.14: 102930476-102930476 on Build GRCh38
rs1036601969 Chr.14: 102930423-102930423 on Build GRCh38
rs987148709 Chr.14: 102930484-102930484 on Build GRCh38
rs778728963 Chr.14: 102930588-102930588 on Build GRCh38

View Full Product Details