Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70596884-70597157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBN
Entrez Gene ID
258
Gene Name
ameloblastin
Gene Aliases
AI1F
UniGene
Hs.272396
Gene Chromosome Location
Chr.4: 70592258-70607288 on Build GRCh38
Transcripts
NM_016519.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1486069 c.96G>T p.Q32H Chr.4: 70597010-70597010 on Build GRCh38
3605482 c.91C>T p.P31S Chr.4: 70597005-70597005 on Build GRCh38
5432194 c.95A>T p.Q32L Chr.4: 70597009-70597009 on Build GRCh38
1753815 c.122G>A p.S41N Chr.4: 70597036-70597036 on Build GRCh38
69628 c.112G>A p.G38S Chr.4: 70597026-70597026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908571877 Chr.4: 70596948-70596948 on Build GRCh38
rs185154087 Chr.4: 70596992-70596992 on Build GRCh38
rs766326078 Chr.4: 70597086-70597086 on Build GRCh38
rs151283081 Chr.4: 70596958-70596958 on Build GRCh38
rs752814924 Chr.4: 70596983-70596983 on Build GRCh38
rs551865058 Chr.4: 70596918-70596918 on Build GRCh38
rs767464468 Chr.4: 70597071-70597071 on Build GRCh38
rs774062263 Chr.4: 70597053-70597053 on Build GRCh38
rs74949118 Chr.4: 70597040-70597040 on Build GRCh38
rs546778141 Chr.4: 70597036-70597036 on Build GRCh38
rs939289963 Chr.4: 70597026-70597026 on Build GRCh38
rs539114576 Chr.4: 70597121-70597121 on Build GRCh38
rs1010641564 Chr.4: 70596908-70596908 on Build GRCh38
rs113127264 Chr.4: 70596966-70596966 on Build GRCh38
rs955805307 Chr.4: 70597047-70597047 on Build GRCh38
rs747166600 Chr.4: 70597013-70597013 on Build GRCh38
rs753659960 Chr.4: 70597100-70597100 on Build GRCh38
rs1022412676 Chr.4: 70596910-70596910 on Build GRCh38
rs371993910 Chr.4: 70596929-70596929 on Build GRCh38
rs767995404 Chr.4: 70597037-70597037 on Build GRCh38
rs533260858 Chr.4: 70596911-70596911 on Build GRCh38
rs761521791 Chr.4: 70597056-70597056 on Build GRCh38
rs771316013 Chr.4: 70597018-70597018 on Build GRCh38
rs571819924 Chr.4: 70597074-70597074 on Build GRCh38
rs982761389 Chr.4: 70596943-70596943 on Build GRCh38
rs377158807 Chr.4: 70596933-70596933 on Build GRCh38
rs1052039914 Chr.4: 70597028-70597028 on Build GRCh38
rs770257329 Chr.4: 70597035-70597035 on Build GRCh38
rs765076338 Chr.4: 70597025-70597025 on Build GRCh38
rs955532764 Chr.4: 70596934-70596934 on Build GRCh38
rs199582898 Chr.4: 70596964-70596964 on Build GRCh38
rs778896618 Chr.4: 70596972-70596972 on Build GRCh38
rs756094947 Chr.4: 70596970-70596970 on Build GRCh38
rs375499565 Chr.4: 70596979-70596979 on Build GRCh38
rs1010979678 Chr.4: 70596939-70596939 on Build GRCh38
rs774984989 Chr.4: 70596966-70596967 on Build GRCh38
rs760758795 Chr.4: 70597081-70597081 on Build GRCh38
rs781488282 Chr.4: 70597024-70597024 on Build GRCh38
rs778067258 Chr.4: 70596993-70596993 on Build GRCh38

View Full Product Details