Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 770352-770625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385356 c.602G>T p.R201I Chr.17: 770449-770449 on Build GRCh38
3820398 c.578T>G p.V193G Chr.17: 770473-770473 on Build GRCh38
5018413 c.622delC p.Q208fs*7 Chr.17: 770429-770429 on Build GRCh38
5722508 c.673G>A p.E225K Chr.17: 770423-770423 on Build GRCh38
4785350 c.647G>T p.R216I Chr.17: 770449-770449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780552852 Chr.17: 770454-770455 on Build GRCh38
rs148496651 Chr.17: 770429-770429 on Build GRCh38
rs751467764 Chr.17: 770465-770465 on Build GRCh38
rs528048808 Chr.17: 770587-770587 on Build GRCh38
rs748952108 Chr.17: 770433-770434 on Build GRCh38
rs746316074 Chr.17: 770539-770539 on Build GRCh38
rs760114727 Chr.17: 770576-770576 on Build GRCh38
rs746961096 Chr.17: 770383-770383 on Build GRCh38
rs111655992 Chr.17: 770413-770413 on Build GRCh38
rs778806817 Chr.17: 770514-770514 on Build GRCh38
rs764860744 Chr.17: 770419-770419 on Build GRCh38
rs760928077 Chr.17: 770542-770542 on Build GRCh38
rs772505963 Chr.17: 770436-770451 on Build GRCh38
rs769097307 Chr.17: 770453-770453 on Build GRCh38
rs780872377 Chr.17: 770386-770386 on Build GRCh38
rs769903260 Chr.17: 770392-770392 on Build GRCh38
rs779142890 Chr.17: 770523-770523 on Build GRCh38
rs142803071 Chr.17: 770494-770494 on Build GRCh38
rs947795789 Chr.17: 770570-770570 on Build GRCh38
rs887429724 Chr.17: 770584-770584 on Build GRCh38
rs115670184 Chr.17: 770486-770486 on Build GRCh38
rs189223792 Chr.17: 770556-770556 on Build GRCh38
rs757876313 Chr.17: 770442-770442 on Build GRCh38
rs948184493 Chr.17: 770538-770538 on Build GRCh38
rs747473177 Chr.17: 770471-770472 on Build GRCh38
rs542652313 Chr.17: 770559-770559 on Build GRCh38
rs780975018 Chr.17: 770472-770472 on Build GRCh38
rs560711423 Chr.17: 770572-770572 on Build GRCh38
rs748345683 Chr.17: 770387-770387 on Build GRCh38
rs767861610 Chr.17: 770407-770407 on Build GRCh38
rs940934181 Chr.17: 770560-770560 on Build GRCh38
rs947438639 Chr.17: 770516-770516 on Build GRCh38
rs750127179 Chr.17: 770421-770421 on Build GRCh38
rs896360290 Chr.17: 770562-770562 on Build GRCh38
rs564071203 Chr.17: 770400-770400 on Build GRCh38
rs113698586 Chr.17: 770461-770461 on Build GRCh38
rs375742621 Chr.17: 770521-770521 on Build GRCh38
rs376272824 Chr.17: 770464-770464 on Build GRCh38
rs769051190 Chr.17: 770551-770551 on Build GRCh38
rs777525022 Chr.17: 770432-770434 on Build GRCh38
rs977763463 Chr.17: 770433-770433 on Build GRCh38
rs774955627 Chr.17: 770401-770401 on Build GRCh38
rs769013224 Chr.17: 770488-770488 on Build GRCh38
rs867091493 Chr.17: 770415-770415 on Build GRCh38
rs373606114 Chr.17: 770522-770522 on Build GRCh38
rs773626869 Chr.17: 770395-770395 on Build GRCh38
rs752123389 Chr.17: 770389-770389 on Build GRCh38
rs754885801 Chr.17: 770466-770466 on Build GRCh38
rs775718133 Chr.17: 770416-770416 on Build GRCh38
rs756022412 Chr.17: 770487-770487 on Build GRCh38
rs759980711 Chr.17: 770404-770404 on Build GRCh38
rs878965532 Chr.17: 770437-770437 on Build GRCh38
rs772456804 Chr.17: 770540-770540 on Build GRCh38
rs755669500 Chr.17: 770396-770396 on Build GRCh38
rs754764583 Chr.17: 770384-770384 on Build GRCh38
rs201697268 Chr.17: 770511-770511 on Build GRCh38
rs749550027 Chr.17: 770507-770507 on Build GRCh38

View Full Product Details