Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47260563-47260804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756982 c.384T>G p.R128R Chr.19: 47260761-47260761 on Build GRCh38
4849911 c.422C>T p.S141F Chr.19: 47260799-47260799 on Build GRCh38
4802115 c.329G>T p.R110L Chr.19: 47260706-47260706 on Build GRCh38
1238461 c.319C>T p.R107* Chr.19: 47260696-47260696 on Build GRCh38
5981572 c.283A>T p.T95S Chr.19: 47260660-47260660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534271315 Chr.19: 47260637-47260637 on Build GRCh38
rs759837994 Chr.19: 47260708-47260708 on Build GRCh38
rs775822299 Chr.19: 47260717-47260717 on Build GRCh38
rs747283701 Chr.19: 47260726-47260726 on Build GRCh38
rs977355900 Chr.19: 47260644-47260644 on Build GRCh38
rs374140233 Chr.19: 47260705-47260705 on Build GRCh38
rs1034930202 Chr.19: 47260601-47260601 on Build GRCh38
rs540587094 Chr.19: 47260759-47260759 on Build GRCh38
rs779041095 Chr.19: 47260781-47260781 on Build GRCh38
rs778263022 Chr.19: 47260584-47260584 on Build GRCh38
rs371400936 Chr.19: 47260725-47260725 on Build GRCh38
rs368381443 Chr.19: 47260706-47260706 on Build GRCh38
rs767997819 Chr.19: 47260670-47260670 on Build GRCh38
rs778877012 Chr.19: 47260633-47260633 on Build GRCh38
rs749365757 Chr.19: 47260704-47260704 on Build GRCh38
rs201968066 Chr.19: 47260631-47260631 on Build GRCh38
rs201682360 Chr.19: 47260686-47260686 on Build GRCh38
rs769576487 Chr.19: 47260739-47260744 on Build GRCh38
rs773494947 Chr.19: 47260594-47260594 on Build GRCh38
rs200713204 Chr.19: 47260651-47260651 on Build GRCh38
rs774665624 Chr.19: 47260663-47260663 on Build GRCh38
rs761267628 Chr.19: 47260718-47260718 on Build GRCh38
rs765361023 Chr.19: 47260592-47260592 on Build GRCh38
rs572199090 Chr.19: 47260765-47260765 on Build GRCh38
rs749356459 Chr.19: 47260772-47260772 on Build GRCh38
rs370729042 Chr.19: 47260703-47260703 on Build GRCh38
rs766756796 Chr.19: 47260597-47260597 on Build GRCh38
rs760039788 Chr.19: 47260681-47260681 on Build GRCh38
rs992420386 Chr.19: 47260738-47260739 on Build GRCh38
rs376079362 Chr.19: 47260697-47260697 on Build GRCh38
rs762044382 Chr.19: 47260774-47260774 on Build GRCh38
rs763156904 Chr.19: 47260595-47260595 on Build GRCh38
rs772108651 Chr.19: 47260635-47260635 on Build GRCh38
rs750101021 Chr.19: 47260739-47260739 on Build GRCh38
rs759960613 Chr.19: 47260669-47260669 on Build GRCh38
rs764561669 Chr.19: 47260726-47260726 on Build GRCh38
rs531794665 Chr.19: 47260613-47260613 on Build GRCh38
rs563018612 Chr.19: 47260590-47260590 on Build GRCh38
rs200754420 Chr.19: 47260692-47260692 on Build GRCh38
rs775539337 Chr.19: 47260709-47260709 on Build GRCh38
rs758222531 Chr.19: 47260748-47260748 on Build GRCh38
rs370298634 Chr.19: 47260610-47260610 on Build GRCh38
rs751356243 Chr.19: 47260764-47260764 on Build GRCh38
rs375104786 Chr.19: 47260630-47260630 on Build GRCh38
rs909191977 Chr.19: 47260746-47260746 on Build GRCh38
rs763256073 Chr.19: 47260658-47260658 on Build GRCh38
rs143218686 Chr.19: 47260760-47260760 on Build GRCh38
rs752012649 Chr.19: 47260609-47260609 on Build GRCh38
rs1049188328 Chr.19: 47260628-47260628 on Build GRCh38
rs551520953 Chr.19: 47260624-47260624 on Build GRCh38
rs929258300 Chr.19: 47260616-47260616 on Build GRCh38
rs772431480 Chr.19: 47260714-47260714 on Build GRCh38
rs1024433069 Chr.19: 47260780-47260780 on Build GRCh38
rs779784977 Chr.19: 47260693-47260693 on Build GRCh38
rs763692575 Chr.19: 47260683-47260683 on Build GRCh38
rs775245963 Chr.19: 47260762-47260776 on Build GRCh38
rs374663536 Chr.19: 47260738-47260738 on Build GRCh38
rs753262442 Chr.19: 47260678-47260678 on Build GRCh38
rs755710031 Chr.19: 47260623-47260623 on Build GRCh38
rs745835993 Chr.19: 47260585-47260585 on Build GRCh38
rs769710715 Chr.19: 47260698-47260698 on Build GRCh38
rs761485205 Chr.19: 47260728-47260728 on Build GRCh38
rs556714252 Chr.19: 47260687-47260687 on Build GRCh38
rs768050798 Chr.19: 47260611-47260611 on Build GRCh38
rs745709397 Chr.19: 47260634-47260634 on Build GRCh38
rs762669943 Chr.19: 47260767-47260768 on Build GRCh38
rs781056381 Chr.19: 47260766-47260766 on Build GRCh38
rs567588231 Chr.19: 47260660-47260660 on Build GRCh38
rs750242270 Chr.19: 47260690-47260690 on Build GRCh38
rs768811346 Chr.19: 47260586-47260586 on Build GRCh38
rs200154609 Chr.19: 47260657-47260657 on Build GRCh38
rs759371866 Chr.19: 47260622-47260622 on Build GRCh38
rs995491239 Chr.19: 47260751-47260751 on Build GRCh38
rs780211130 Chr.19: 47260642-47260642 on Build GRCh38

View Full Product Details