Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 80272956-80273181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDHB
Entrez Gene ID
594
Gene Name
branched chain keto acid dehydrogenase E1 subunit beta
Gene Aliases
BCKDE1B, BCKDH E1-beta, E1B
UniGene
Hs.654441
Gene Chromosome Location
Chr.6: 80106610-80469088 on Build GRCh38
Transcripts
NM_000056.4 NM_001318975.1 NM_183050.3 NR_134945.1 XM_005248756.4 XM_011536023.2 XM_011536024.2 XM_011536025.2 XM_011536026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
270688 c.971G>A p.R324Q Chr.6: 80273154-80273154 on Build GRCh38
1446101 c.970C>A p.R324R Chr.6: 80273153-80273153 on Build GRCh38
301754 c.996C>A p.P332P Chr.6: 80273179-80273179 on Build GRCh38
1672585 c.970C>T p.R324* Chr.6: 80273153-80273153 on Build GRCh38
5421614 c.965C>T p.T322I Chr.6: 80273148-80273148 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759198446 Chr.6: 80273109-80273109 on Build GRCh38
rs779150907 Chr.6: 80273154-80273154 on Build GRCh38
rs996013555 Chr.6: 80273048-80273048 on Build GRCh38
rs767009413 Chr.6: 80273116-80273116 on Build GRCh38
rs931968192 Chr.6: 80273081-80273081 on Build GRCh38
rs890103324 Chr.6: 80273116-80273117 on Build GRCh38
rs765905155 Chr.6: 80273087-80273087 on Build GRCh38
rs398124604 Chr.6: 80273157-80273157 on Build GRCh38
rs372013960 Chr.6: 80273144-80273144 on Build GRCh38
rs973553024 Chr.6: 80273052-80273052 on Build GRCh38
rs764707388 Chr.6: 80273152-80273152 on Build GRCh38
rs398124602 Chr.6: 80273134-80273134 on Build GRCh38
rs559388556 Chr.6: 80272997-80272997 on Build GRCh38
rs753170463 Chr.6: 80273143-80273143 on Build GRCh38
rs565119070 Chr.6: 80273096-80273096 on Build GRCh38
rs1050398834 Chr.6: 80273085-80273085 on Build GRCh38
rs528490227 Chr.6: 80273006-80273006 on Build GRCh38
rs398124603 Chr.6: 80273153-80273153 on Build GRCh38
rs9341811 Chr.6: 80272984-80272984 on Build GRCh38
rs748534284 Chr.6: 80273102-80273102 on Build GRCh38
rs869312131 Chr.6: 80273147-80273147 on Build GRCh38
rs545413403 Chr.6: 80273069-80273069 on Build GRCh38

View Full Product Details