Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67507986-67508257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK2AP2
Entrez Gene ID
10263
Gene Name
cyclin dependent kinase 2 associated protein 2
Gene Aliases
DOC-1R, p14
UniGene
Hs.523835
Gene Chromosome Location
Chr.11: 67506490-67508728 on Build GRCh38
Transcripts
NM_001271849.1 NM_005851.4 NR_073484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499796 c.54C>T p.T18T Chr.11: 67508029-67508029 on Build GRCh38
6563256 c.25G>A p.A9T Chr.11: 67508058-67508058 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs889035366 Chr.11: 67508169-67508169 on Build GRCh38
rs546802271 Chr.11: 67508144-67508144 on Build GRCh38
rs577841821 Chr.11: 67508089-67508089 on Build GRCh38
rs1051592757 Chr.11: 67508165-67508165 on Build GRCh38
rs770485066 Chr.11: 67508100-67508100 on Build GRCh38
rs769616262 Chr.11: 67508115-67508115 on Build GRCh38
rs548857667 Chr.11: 67508077-67508077 on Build GRCh38
rs566031302 Chr.11: 67508125-67508125 on Build GRCh38
rs11548606 Chr.11: 67508194-67508194 on Build GRCh38
rs147979185 Chr.11: 67508161-67508161 on Build GRCh38
rs765859666 Chr.11: 67508125-67508125 on Build GRCh38
rs759621854 Chr.11: 67508014-67508014 on Build GRCh38
rs538871541 Chr.11: 67508175-67508175 on Build GRCh38
rs776403398 Chr.11: 67508104-67508104 on Build GRCh38
rs917056627 Chr.11: 67508179-67508179 on Build GRCh38
rs1038360871 Chr.11: 67508238-67508238 on Build GRCh38
rs997396922 Chr.11: 67508162-67508162 on Build GRCh38
rs1029816316 Chr.11: 67508087-67508087 on Build GRCh38
rs775293601 Chr.11: 67508118-67508118 on Build GRCh38
rs1044664635 Chr.11: 67508202-67508202 on Build GRCh38
rs560982282 Chr.11: 67508107-67508107 on Build GRCh38
rs530762126 Chr.11: 67508045-67508045 on Build GRCh38
rs1036173242 Chr.11: 67508148-67508148 on Build GRCh38
rs1007671902 Chr.11: 67508172-67508172 on Build GRCh38
rs765388352 Chr.11: 67508070-67508070 on Build GRCh38
rs528466504 Chr.11: 67508114-67508114 on Build GRCh38
rs777361847 Chr.11: 67508065-67508065 on Build GRCh38
rs748629135 Chr.11: 67508039-67508039 on Build GRCh38
rs1009296601 Chr.11: 67508237-67508237 on Build GRCh38
rs909319613 Chr.11: 67508139-67508139 on Build GRCh38
rs779536080 Chr.11: 67508024-67508024 on Build GRCh38
rs754144320 Chr.11: 67508007-67508007 on Build GRCh38
rs941357607 Chr.11: 67508213-67508213 on Build GRCh38
rs755423555 Chr.11: 67508013-67508013 on Build GRCh38
rs758999590 Chr.11: 67508047-67508047 on Build GRCh38
rs1007529988 Chr.11: 67508142-67508142 on Build GRCh38
rs536983449 Chr.11: 67508217-67508217 on Build GRCh38
rs937297302 Chr.11: 67508072-67508072 on Build GRCh38
rs928659152 Chr.11: 67508020-67508020 on Build GRCh38
rs551145947 Chr.11: 67508178-67508178 on Build GRCh38
rs927238416 Chr.11: 67508205-67508205 on Build GRCh38

View Full Product Details