Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28716166-28716439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5409381 c.175G>T p.E59* Chr.8: 28716234-28716234 on Build GRCh38
6262622 c.207G>T p.V69V Chr.8: 28716266-28716266 on Build GRCh38
1098914 c.221G>A p.C74Y Chr.8: 28716280-28716280 on Build GRCh38
305658 c.169G>T p.A57S Chr.8: 28716228-28716228 on Build GRCh38
5109927 c.266T>C p.V89A Chr.8: 28716325-28716325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762082642 Chr.8: 28716233-28716233 on Build GRCh38
rs369407692 Chr.8: 28716256-28716256 on Build GRCh38
rs769813480 Chr.8: 28716372-28716372 on Build GRCh38
rs773454046 Chr.8: 28716374-28716374 on Build GRCh38
rs910806899 Chr.8: 28716194-28716194 on Build GRCh38
rs372678243 Chr.8: 28716212-28716212 on Build GRCh38
rs142901670 Chr.8: 28716297-28716297 on Build GRCh38
rs746203687 Chr.8: 28716265-28716265 on Build GRCh38
rs764082441 Chr.8: 28716403-28716403 on Build GRCh38
rs745551307 Chr.8: 28716328-28716328 on Build GRCh38
rs993707572 Chr.8: 28716359-28716359 on Build GRCh38
rs778434439 Chr.8: 28716264-28716264 on Build GRCh38
rs1046622436 Chr.8: 28716229-28716229 on Build GRCh38
rs776040229 Chr.8: 28716273-28716273 on Build GRCh38
rs748933869 Chr.8: 28716258-28716258 on Build GRCh38
rs766037251 Chr.8: 28716323-28716323 on Build GRCh38
rs373430705 Chr.8: 28716246-28716246 on Build GRCh38
rs1012943161 Chr.8: 28716290-28716290 on Build GRCh38
rs1027379756 Chr.8: 28716365-28716365 on Build GRCh38
rs376529517 Chr.8: 28716247-28716247 on Build GRCh38
rs776636386 Chr.8: 28716282-28716282 on Build GRCh38
rs754765415 Chr.8: 28716243-28716243 on Build GRCh38
rs772342569 Chr.8: 28716272-28716272 on Build GRCh38
rs200105783 Chr.8: 28716293-28716293 on Build GRCh38
rs761888477 Chr.8: 28716221-28716222 on Build GRCh38
rs764137887 Chr.8: 28716312-28716312 on Build GRCh38
rs772563405 Chr.8: 28716190-28716190 on Build GRCh38
rs777157868 Chr.8: 28716344-28716344 on Build GRCh38
rs760693109 Chr.8: 28716198-28716198 on Build GRCh38
rs763152974 Chr.8: 28716375-28716375 on Build GRCh38
rs746445288 Chr.8: 28716348-28716349 on Build GRCh38
rs907192576 Chr.8: 28716299-28716299 on Build GRCh38
rs747499791 Chr.8: 28716338-28716338 on Build GRCh38
rs1035328996 Chr.8: 28716320-28716320 on Build GRCh38
rs375815743 Chr.8: 28716200-28716200 on Build GRCh38
rs199979119 Chr.8: 28716381-28716381 on Build GRCh38
rs370320793 Chr.8: 28716242-28716242 on Build GRCh38
rs781136425 Chr.8: 28716245-28716245 on Build GRCh38
rs780460600 Chr.8: 28716331-28716331 on Build GRCh38
rs759352865 Chr.8: 28716295-28716295 on Build GRCh38
rs146089300 Chr.8: 28716380-28716380 on Build GRCh38
rs753849077 Chr.8: 28716412-28716412 on Build GRCh38
rs376405458 Chr.8: 28716281-28716281 on Build GRCh38
rs374083446 Chr.8: 28716294-28716294 on Build GRCh38
rs747551309 Chr.8: 28716278-28716278 on Build GRCh38
rs538331636 Chr.8: 28716401-28716401 on Build GRCh38
rs369033887 Chr.8: 28716238-28716238 on Build GRCh38
rs756180786 Chr.8: 28716302-28716302 on Build GRCh38
rs760548680 Chr.8: 28716402-28716402 on Build GRCh38
rs189382426 Chr.8: 28716357-28716357 on Build GRCh38
rs377363064 Chr.8: 28716339-28716339 on Build GRCh38
rs774502059 Chr.8: 28716209-28716209 on Build GRCh38
rs370448870 Chr.8: 28716356-28716356 on Build GRCh38
rs753330593 Chr.8: 28716315-28716315 on Build GRCh38
rs754024716 Chr.8: 28716219-28716219 on Build GRCh38
rs756747852 Chr.8: 28716322-28716322 on Build GRCh38
rs967044101 Chr.8: 28716355-28716355 on Build GRCh38
rs150466027 Chr.8: 28716262-28716262 on Build GRCh38
rs141531460 Chr.8: 28716197-28716197 on Build GRCh38
rs767256866 Chr.8: 28716296-28716296 on Build GRCh38

View Full Product Details