Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20525737-20525976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYL1
Entrez Gene ID
51084
Gene Name
crystallin lambda 1
Gene Aliases
GDH, HEL30, lambda-CRY
UniGene
Hs.370703
Gene Chromosome Location
Chr.13: 20403667-20525873 on Build GRCh38
Transcripts
NM_015974.2 XM_005266415.3 XM_005266416.4 XM_017020624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376905 c.20G>T p.G7V Chr.13: 20525775-20525775 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999672980 Chr.13: 20525825-20525826 on Build GRCh38
rs902743198 Chr.13: 20525928-20525929 on Build GRCh38
rs768761618 Chr.13: 20525796-20525796 on Build GRCh38
rs887907381 Chr.13: 20525873-20525873 on Build GRCh38
rs572427202 Chr.13: 20525787-20525787 on Build GRCh38
rs1023632034 Chr.13: 20525851-20525851 on Build GRCh38
rs1049437474 Chr.13: 20525803-20525803 on Build GRCh38
rs761924244 Chr.13: 20525828-20525828 on Build GRCh38
rs925200785 Chr.13: 20525950-20525950 on Build GRCh38
rs898060061 Chr.13: 20525856-20525856 on Build GRCh38
rs375294912 Chr.13: 20525843-20525843 on Build GRCh38
rs1035534593 Chr.13: 20525839-20525839 on Build GRCh38
rs1012666930 Chr.13: 20525846-20525846 on Build GRCh38
rs912348589 Chr.13: 20525804-20525804 on Build GRCh38
rs1049797363 Chr.13: 20525864-20525864 on Build GRCh38
rs774572461 Chr.13: 20525814-20525814 on Build GRCh38
rs1022444857 Chr.13: 20525812-20525812 on Build GRCh38
rs1011111227 Chr.13: 20525810-20525810 on Build GRCh38
rs1037175661 Chr.13: 20525774-20525774 on Build GRCh38
rs1056599351 Chr.13: 20525959-20525959 on Build GRCh38
rs910870754 Chr.13: 20525879-20525879 on Build GRCh38
rs993736485 Chr.13: 20525863-20525863 on Build GRCh38
rs979769131 Chr.13: 20525770-20525770 on Build GRCh38
rs192552300 Chr.13: 20525915-20525915 on Build GRCh38
rs879025449 Chr.13: 20525832-20525832 on Build GRCh38
rs554211019 Chr.13: 20525784-20525784 on Build GRCh38
rs1033761722 Chr.13: 20525878-20525878 on Build GRCh38
rs984019253 Chr.13: 20525857-20525857 on Build GRCh38
rs556050237 Chr.13: 20525925-20525925 on Build GRCh38
rs960804312 Chr.13: 20525880-20525880 on Build GRCh38
rs796865876 Chr.13: 20525779-20525779 on Build GRCh38
rs902670234 Chr.13: 20525818-20525818 on Build GRCh38
rs188891229 Chr.13: 20525875-20525875 on Build GRCh38
rs781407016 Chr.13: 20525777-20525777 on Build GRCh38
rs909744515 Chr.13: 20525868-20525868 on Build GRCh38
rs746419888 Chr.13: 20525783-20525783 on Build GRCh38
rs1013990402 Chr.13: 20525898-20525898 on Build GRCh38
rs967130156 Chr.13: 20525924-20525924 on Build GRCh38
rs868063316 Chr.13: 20525935-20525935 on Build GRCh38
rs140651902 Chr.13: 20525795-20525795 on Build GRCh38

View Full Product Details