Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43353824-43354051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD177
Entrez Gene ID
57126
Gene Name
CD177 molecule
Gene Aliases
HNA-2a, HNA2A, NB1, NB1 GP, PRV-1, PRV1
UniGene
Hs.232165
Gene Chromosome Location
Chr.19: 43353659-43366075 on Build GRCh38
Transcripts
NM_020406.3 XM_017027021.1 XM_017027022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3892774 c.125G>A p.W42* Chr.19: 43353925-43353925 on Build GRCh38
5678177 c.109G>T p.D37Y Chr.19: 43353909-43353909 on Build GRCh38
7305153 c.153C>T p.S51S Chr.19: 43353953-43353953 on Build GRCh38
5678176 c.109G>T p.D37Y Chr.19: 43353909-43353909 on Build GRCh38
4188652 c.116C>T p.P39L Chr.19: 43353916-43353916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376471344 Chr.19: 43353856-43353856 on Build GRCh38
rs749447405 Chr.19: 43354016-43354016 on Build GRCh38
rs745611109 Chr.19: 43353959-43353959 on Build GRCh38
rs778001389 Chr.19: 43354006-43354006 on Build GRCh38
rs766286384 Chr.19: 43353952-43353952 on Build GRCh38
rs756155144 Chr.19: 43354002-43354002 on Build GRCh38
rs781707883 Chr.19: 43353995-43353995 on Build GRCh38
rs929905307 Chr.19: 43353969-43353969 on Build GRCh38
rs1025318990 Chr.19: 43354028-43354028 on Build GRCh38
rs372638050 Chr.19: 43353903-43353903 on Build GRCh38
rs779990458 Chr.19: 43354019-43354019 on Build GRCh38
rs750077398 Chr.19: 43353983-43353983 on Build GRCh38
rs760839478 Chr.19: 43353919-43353919 on Build GRCh38
rs45553433 Chr.19: 43353892-43353892 on Build GRCh38
rs776789402 Chr.19: 43353874-43353874 on Build GRCh38
rs553076298 Chr.19: 43354027-43354027 on Build GRCh38
rs746854959 Chr.19: 43354022-43354022 on Build GRCh38
rs556902433 Chr.19: 43353972-43353972 on Build GRCh38
rs552335761 Chr.19: 43353899-43353899 on Build GRCh38
rs781221581 Chr.19: 43353941-43353941 on Build GRCh38
rs548201107 Chr.19: 43353925-43353925 on Build GRCh38
rs747027356 Chr.19: 43353846-43353846 on Build GRCh38
rs750739565 Chr.19: 43353918-43353918 on Build GRCh38
rs375804751 Chr.19: 43353943-43353943 on Build GRCh38
rs375460393 Chr.19: 43353909-43353909 on Build GRCh38
rs576931329 Chr.19: 43353987-43353987 on Build GRCh38
rs372486994 Chr.19: 43353932-43353932 on Build GRCh38
rs771223359 Chr.19: 43354017-43354017 on Build GRCh38
rs372882731 Chr.19: 43353975-43353975 on Build GRCh38
rs763675091 Chr.19: 43353907-43353907 on Build GRCh38
rs752085844 Chr.19: 43353930-43353930 on Build GRCh38
rs764874527 Chr.19: 43353982-43353982 on Build GRCh38
rs755244771 Chr.19: 43353997-43353997 on Build GRCh38
rs767433966 Chr.19: 43353896-43353896 on Build GRCh38
rs768758511 Chr.19: 43353873-43353873 on Build GRCh38
rs201150439 Chr.19: 43353968-43353968 on Build GRCh38
rs1033013795 Chr.19: 43354013-43354013 on Build GRCh38
rs751964781 Chr.19: 43353993-43353993 on Build GRCh38
rs1023453294 Chr.19: 43353988-43353988 on Build GRCh38
rs761418116 Chr.19: 43353980-43353980 on Build GRCh38
rs748352348 Chr.19: 43353881-43353881 on Build GRCh38
rs45571738 Chr.19: 43353914-43353914 on Build GRCh38
rs534652679 Chr.19: 43353919-43353919 on Build GRCh38
rs140028442 Chr.19: 43354001-43354001 on Build GRCh38
rs753341493 Chr.19: 43353951-43353951 on Build GRCh38
rs766634373 Chr.19: 43353989-43353989 on Build GRCh38
rs568275482 Chr.19: 43353953-43353953 on Build GRCh38
rs754133534 Chr.19: 43353956-43353956 on Build GRCh38
rs1038462274 Chr.19: 43353940-43353940 on Build GRCh38
rs759308338 Chr.19: 43353894-43353894 on Build GRCh38
rs777873188 Chr.19: 43353954-43353954 on Build GRCh38

View Full Product Details