Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186162830-186163055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149A
Entrez Gene ID
25854
Gene Name
family with sequence similarity 149 member A
Gene Aliases
MST119, MSTP119
UniGene
Hs.744961
Gene Chromosome Location
Chr.4: 186104798-186172663 on Build GRCh38
Transcripts
NM_001006655.2 NM_015398.2 XM_005262911.1 XM_005262912.1 XM_005262913.1 XM_011531834.1 XM_011531835.2 XM_011531836.1 XM_017007996.1 XM_017007997.1 XM_017007998.1 XM_017007999.1 XM_017008000.1 XM_017008001.1 XM_017008002.1 XM_017008003.1 XM_017008004.1 XM_017008005.1 XM_017008006.1 XM_017008007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557348 c.1603A>T p.I535F Chr.4: 186162845-186162845 on Build GRCh38
6166876 c.763A>C p.M255L Chr.4: 186162878-186162878 on Build GRCh38
1053890 c.795G>A p.P265P Chr.4: 186162910-186162910 on Build GRCh38
7367073 c.1627T>G p.Y543D Chr.4: 186162869-186162869 on Build GRCh38
4158948 c.791A>C p.K264T Chr.4: 186162906-186162906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759704535 Chr.4: 186162859-186162859 on Build GRCh38
rs377734857 Chr.4: 186162945-186162945 on Build GRCh38
rs765813557 Chr.4: 186162974-186162974 on Build GRCh38
rs1009916117 Chr.4: 186162996-186162996 on Build GRCh38
rs553556070 Chr.4: 186162892-186162892 on Build GRCh38
rs201887086 Chr.4: 186162973-186162973 on Build GRCh38
rs1036050339 Chr.4: 186162891-186162891 on Build GRCh38
rs559042193 Chr.4: 186162864-186162864 on Build GRCh38
rs75674583 Chr.4: 186163025-186163025 on Build GRCh38
rs142363212 Chr.4: 186162932-186162932 on Build GRCh38
rs918867886 Chr.4: 186163002-186163002 on Build GRCh38
rs775749597 Chr.4: 186162952-186162952 on Build GRCh38
rs146121160 Chr.4: 186162946-186162946 on Build GRCh38
rs770818438 Chr.4: 186162863-186162864 on Build GRCh38
rs779900401 Chr.4: 186162917-186162917 on Build GRCh38
rs940252411 Chr.4: 186162887-186162887 on Build GRCh38
rs77537358 Chr.4: 186162883-186162883 on Build GRCh38
rs942800835 Chr.4: 186163028-186163028 on Build GRCh38
rs538796693 Chr.4: 186163019-186163019 on Build GRCh38
rs1000736356 Chr.4: 186162909-186162909 on Build GRCh38
rs138445627 Chr.4: 186162886-186162886 on Build GRCh38
rs757955183 Chr.4: 186162910-186162910 on Build GRCh38
rs776685468 Chr.4: 186162941-186162942 on Build GRCh38
rs530481027 Chr.4: 186162930-186162930 on Build GRCh38
rs530341646 Chr.4: 186162955-186162955 on Build GRCh38
rs370171509 Chr.4: 186162953-186162953 on Build GRCh38
rs545080144 Chr.4: 186162878-186162878 on Build GRCh38
rs754410805 Chr.4: 186162880-186162880 on Build GRCh38
rs75877362 Chr.4: 186163003-186163003 on Build GRCh38
rs768345782 Chr.4: 186162920-186162920 on Build GRCh38
rs754313295 Chr.4: 186162958-186162958 on Build GRCh38
rs750730149 Chr.4: 186162981-186162981 on Build GRCh38
rs374453509 Chr.4: 186162918-186162918 on Build GRCh38
rs781205660 Chr.4: 186162927-186162927 on Build GRCh38
rs79537217 Chr.4: 186162869-186162869 on Build GRCh38
rs772262578 Chr.4: 186162947-186162947 on Build GRCh38
rs756237721 Chr.4: 186162873-186162873 on Build GRCh38
rs779158731 Chr.4: 186162884-186162884 on Build GRCh38
rs758853926 Chr.4: 186162991-186162991 on Build GRCh38

View Full Product Details