Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6165530-6165775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
UniGene
Hs.487479
Gene Chromosome Location
Chr.7: 6161779-6272611 on Build GRCh38
Transcripts
NM_004227.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4904087 c.700G>A p.E234K Chr.7: 6165562-6165562 on Build GRCh38
6639579 c.868G>A p.D290N Chr.7: 6165766-6165766 on Build GRCh38
4905638 c.670C>T p.P224S Chr.7: 6165592-6165592 on Build GRCh38
1091139 c.972+1G>A p.? Chr.7: 6165544-6165544 on Build GRCh38
6639591 c.709C>T p.R237W Chr.7: 6165553-6165553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900782113 Chr.7: 6165625-6165625 on Build GRCh38
rs569580678 Chr.7: 6165630-6165630 on Build GRCh38
rs765504329 Chr.7: 6165643-6165643 on Build GRCh38
rs374575146 Chr.7: 6165636-6165636 on Build GRCh38
rs774463026 Chr.7: 6165637-6165637 on Build GRCh38
rs757740683 Chr.7: 6165687-6165687 on Build GRCh38
rs377725186 Chr.7: 6165638-6165638 on Build GRCh38
rs756810208 Chr.7: 6165623-6165623 on Build GRCh38
rs746541589 Chr.7: 6165663-6165663 on Build GRCh38
rs938456652 Chr.7: 6165563-6165563 on Build GRCh38
rs983618718 Chr.7: 6165589-6165589 on Build GRCh38
rs376635813 Chr.7: 6165692-6165692 on Build GRCh38
rs766331224 Chr.7: 6165557-6165557 on Build GRCh38
rs769399428 Chr.7: 6165629-6165629 on Build GRCh38
rs554474156 Chr.7: 6165626-6165626 on Build GRCh38
rs747471595 Chr.7: 6165621-6165621 on Build GRCh38
rs776614433 Chr.7: 6165639-6165639 on Build GRCh38
rs755203995 Chr.7: 6165560-6165560 on Build GRCh38
rs755591853 Chr.7: 6165701-6165701 on Build GRCh38
rs373912967 Chr.7: 6165708-6165708 on Build GRCh38
rs759661343 Chr.7: 6165642-6165642 on Build GRCh38
rs369718141 Chr.7: 6165658-6165658 on Build GRCh38
rs752862361 Chr.7: 6165649-6165649 on Build GRCh38
rs758769237 Chr.7: 6165654-6165654 on Build GRCh38
rs758752571 Chr.7: 6165608-6165608 on Build GRCh38
rs370816256 Chr.7: 6165707-6165707 on Build GRCh38
rs1021525922 Chr.7: 6165675-6165675 on Build GRCh38
rs778268461 Chr.7: 6165619-6165619 on Build GRCh38
rs762354199 Chr.7: 6165701-6165701 on Build GRCh38
rs575115053 Chr.7: 6165572-6165572 on Build GRCh38
rs189205553 Chr.7: 6165590-6165590 on Build GRCh38
rs775266968 Chr.7: 6165635-6165635 on Build GRCh38
rs778901451 Chr.7: 6165729-6165729 on Build GRCh38
rs768208726 Chr.7: 6165724-6165725 on Build GRCh38
rs1011417033 Chr.7: 6165664-6165664 on Build GRCh38
rs751897023 Chr.7: 6165667-6165667 on Build GRCh38
rs199620770 Chr.7: 6165651-6165651 on Build GRCh38
rs768596669 Chr.7: 6165725-6165725 on Build GRCh38
rs760840529 Chr.7: 6165553-6165553 on Build GRCh38
rs373659033 Chr.7: 6165691-6165691 on Build GRCh38
rs200366889 Chr.7: 6165627-6165627 on Build GRCh38
rs747945155 Chr.7: 6165740-6165740 on Build GRCh38
rs754113042 Chr.7: 6165558-6165558 on Build GRCh38
rs771974685 Chr.7: 6165743-6165743 on Build GRCh38
rs969973797 Chr.7: 6165689-6165689 on Build GRCh38

View Full Product Details