Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109252452-109252709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
UniGene
Hs.57652
Gene Chromosome Location
Chr.1: 109249705-109275756 on Build GRCh38
Transcripts
NM_001408.2 XM_005270580.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457651 c.2624C>T p.T875M Chr.1: 109252703-109252703 on Build GRCh38
674478 c.2575G>C p.D859H Chr.1: 109252654-109252654 on Build GRCh38
6120121 c.2389G>T p.D797Y Chr.1: 109252468-109252468 on Build GRCh38
1320169 c.2385G>A p.K795K Chr.1: 109252464-109252464 on Build GRCh38
5968718 c.2572G>A p.D858N Chr.1: 109252651-109252651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773358358 Chr.1: 109252644-109252644 on Build GRCh38
rs777270805 Chr.1: 109252511-109252511 on Build GRCh38
rs565216430 Chr.1: 109252653-109252653 on Build GRCh38
rs373425704 Chr.1: 109252480-109252480 on Build GRCh38
rs754886779 Chr.1: 109252579-109252579 on Build GRCh38
rs151136134 Chr.1: 109252611-109252611 on Build GRCh38
rs573729296 Chr.1: 109252513-109252513 on Build GRCh38
rs745566729 Chr.1: 109252685-109252685 on Build GRCh38
rs1026244756 Chr.1: 109252622-109252622 on Build GRCh38
rs750573929 Chr.1: 109252561-109252561 on Build GRCh38
rs749644861 Chr.1: 109252628-109252628 on Build GRCh38
rs953299138 Chr.1: 109252492-109252492 on Build GRCh38
rs756947114 Chr.1: 109252623-109252623 on Build GRCh38
rs370447679 Chr.1: 109252663-109252663 on Build GRCh38
rs770574783 Chr.1: 109252678-109252679 on Build GRCh38
rs962076401 Chr.1: 109252509-109252509 on Build GRCh38
rs928776423 Chr.1: 109252514-109252514 on Build GRCh38
rs771982823 Chr.1: 109252640-109252640 on Build GRCh38
rs1039838854 Chr.1: 109252597-109252597 on Build GRCh38
rs769134813 Chr.1: 109252536-109252536 on Build GRCh38
rs377135836 Chr.1: 109252661-109252661 on Build GRCh38
rs774201169 Chr.1: 109252680-109252681 on Build GRCh38
rs762667468 Chr.1: 109252686-109252686 on Build GRCh38
rs370616613 Chr.1: 109252651-109252651 on Build GRCh38
rs142987028 Chr.1: 109252497-109252497 on Build GRCh38
rs554845360 Chr.1: 109252494-109252494 on Build GRCh38
rs376119081 Chr.1: 109252501-109252501 on Build GRCh38
rs991646398 Chr.1: 109252565-109252565 on Build GRCh38
rs35689464 Chr.1: 109252513-109252514 on Build GRCh38
rs201283895 Chr.1: 109252654-109252654 on Build GRCh38
rs757931859 Chr.1: 109252603-109252603 on Build GRCh38
rs201340375 Chr.1: 109252570-109252570 on Build GRCh38
rs370538970 Chr.1: 109252503-109252503 on Build GRCh38
rs774901050 Chr.1: 109252548-109252548 on Build GRCh38
rs376437956 Chr.1: 109252578-109252578 on Build GRCh38
rs753655956 Chr.1: 109252495-109252495 on Build GRCh38
rs761847500 Chr.1: 109252553-109252553 on Build GRCh38
rs535934834 Chr.1: 109252572-109252572 on Build GRCh38
rs751067182 Chr.1: 109252668-109252668 on Build GRCh38
rs763391847 Chr.1: 109252529-109252529 on Build GRCh38
rs755819249 Chr.1: 109252473-109252473 on Build GRCh38
rs752733978 Chr.1: 109252587-109252587 on Build GRCh38
rs756780197 Chr.1: 109252670-109252670 on Build GRCh38
rs780762006 Chr.1: 109252624-109252624 on Build GRCh38
rs572525434 Chr.1: 109252621-109252621 on Build GRCh38
rs760728043 Chr.1: 109252650-109252650 on Build GRCh38
rs373423391 Chr.1: 109252557-109252557 on Build GRCh38
rs544567629 Chr.1: 109252630-109252630 on Build GRCh38
rs747656635 Chr.1: 109252500-109252500 on Build GRCh38
rs150411435 Chr.1: 109252662-109252662 on Build GRCh38
rs145105634 Chr.1: 109252525-109252525 on Build GRCh38
rs748687661 Chr.1: 109252635-109252635 on Build GRCh38
rs774815074 Chr.1: 109252632-109252632 on Build GRCh38
rs1006432764 Chr.1: 109252680-109252680 on Build GRCh38
rs901619773 Chr.1: 109252637-109252637 on Build GRCh38
rs746574787 Chr.1: 109252618-109252618 on Build GRCh38
rs765237291 Chr.1: 109252584-109252584 on Build GRCh38
rs143186589 Chr.1: 109252673-109252673 on Build GRCh38
rs760959590 Chr.1: 109252567-109252567 on Build GRCh38

View Full Product Details